Skip to Content
Merck
All Photos(8)

Key Documents

HPA001198

Sigma-Aldrich

Anti-BTK antibody produced in rabbit

enhanced validation

Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-ATK antibody produced in rabbit, Anti-Agammaglobulinemia tyrosine kinase antibody produced in rabbit, Anti-B cell progenitor kinase antibody produced in rabbit, Anti-BPK antibody produced in rabbit, Anti-Bruton tyrosine kinase antibody produced in rabbit, Anti-Tyrosine-protein kinase BTK antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

PAAAPVSTSELKKVVALYDYMPMNANDLQLRKGDEYFILEESNLPWWRARDKNGQEGYIPSNYVTEAEDSIEMYEWYSKHMTRSQAEQLLKQEGKEGGFIVRDSSKAGKYTVSVFAKSTGDPQGVIRHYVVCSTPQSQYYLAEKHL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BTK(695)

Looking for similar products? Visit Product Comparison Guide

Immunogen

Tyrosine-protein kinase BTK recombinant protein epitope signature tag (PrEST)

Application

Anti-BTK antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

BTK (Bruton agammaglobulinemia tyrosine kinase) gene encodes a non-receptor tyrosine kinase that has a crucial role in B lymphocyte development, differentiation and signaling. It is essential for lipopolysaccharide (LPS)-induced as well as Toll-like receptors (TLR2 and TLR4)-induced tumor necrosis factor (TNF) production. Bright, a member of the ARID family of transcription factors, promotes immunoglobulin heavy-chain transcription via association with the transcription factor TFII-I and Btk. Btk interacts with and activates signaling by Toll-like receptors (TLR8 and TLR9) that are involved in the detection of pathogens and activation of host defense. Defects in this gene cause X-linked agammaglobulinaemia (XLA), a humoral immunodeficiency disease.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST78234

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jaya Rajaiya et al.
Molecular and cellular biology, 26(12), 4758-4768 (2006-06-02)
Bright/ARID3a/Dril1, a member of the ARID family of transcription factors, is expressed in a highly regulated fashion in B lymphocytes, where it enhances immunoglobulin transcription three- to sixfold. Recent publications from our lab indicated that functional, but not kinase-inactive, Bruton's
Laia Sadeghi et al.
Cancers, 12(5) (2020-05-07)
Interactions between lymphoma cells and stromal cells play a key role in promoting tumor survival and development of drug resistance. We identified differences in key signaling pathways between the JeKo-1 and REC-1 mantle cell lymphoma (MCL) cell lines, displaying different
Sanghee Lim et al.
Acta neuropathologica communications, 12(1), 56-56 (2024-04-09)
In malignant glioma, cytotoxic drugs are often inhibited from accessing the tumor site due to the blood-tumor barrier (BTB). Ibrutinib, FDA-approved lymphoma agent, inhibits Bruton tyrosine kinase (BTK) and has previously been shown to independently impair aortic endothelial adhesion and
W Yang et al.
Proceedings of the National Academy of Sciences of the United States of America, 94(2), 604-609 (1997-01-21)
Bruton's tyrosine kinase (Btk) is essential for B cell activation, but downstream targets of Btk have not been defined. We now describe a protein, BAP-135, that is associated with Btk in B cells and is a substrate for phosphorylation by
Nicole J Horwood et al.
Journal of immunology (Baltimore, Md. : 1950), 176(6), 3635-3641 (2006-03-07)
Bruton's tyrosine kinase (Btk), the gene mutated in the human immunodeficiency X-linked agammaglobulinemia, is activated by LPS and is required for LPS-induced TNF production. In this study, we have investigated the role of Btk both in signaling via another TLR

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service