Skip to Content
Merck
All Photos(10)

Key Documents

HPA000839

Sigma-Aldrich

Anti-GABRA3 antibody produced in rabbit

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-γ-Aminobutyric-acid receptor subunit α-3 precursor antibody produced in rabbit, Anti-GABA(A) receptor subunit α-3 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

TSHCYMTSLGILFLINILPGTTGQGESRRQEPGDFVKQDIGGLSPKHAPDIPDDSTDNITIFTRILDRLLDGYDNRLRPGLGDAVTEVKTDIYVTSFGPVSDTDMEYTIDVFFRQTWHDE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GABRA3(2556)

General description

γ-aminobutyric acid receptor subunit α-3 is a protein encoded by the GABRA3 gene in humans and is mapped to chromosome X.

Immunogen

γ-Aminobutyric-acid receptor subunit α-3 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

γ-aminobutyric acid receptor subunit α-3 plays an important role in hepatocellular carcinoma (HCC) development and progression. It may act as a promising molecular target for the development of new diagnostic and therapeutic strategies for HCC. Overexpression of GABRA3 is found in lung cancer and rarely in other cancers. It can act as a potential biomarker for diagnosis of lung cancer and the gene receptors may play an important role in cancer differentiation. GABRA3 may be involved in the genetic pathophysiology of unipolar major depressive disorder.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74005

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sara Brin Rosenthal et al.
Hepatology (Baltimore, Md.), 74(2), 667-685 (2021-02-08)
In clinical and experimental NASH, the origin of the scar-forming myofibroblast is the HSC. We used foz/foz mice on a Western diet to characterize in detail the phenotypic changes of HSCs in a NASH model. We examined the single-cell expression
Yan Liu et al.
Pathology oncology research : POR, 15(3), 351-358 (2008-12-03)
The identification of tumor-associated antigens, which are specifically expressed in cancer tissues, is very important for immunotherapy of lung cancer. We have combined the in silico screening and experimental verifying to identify genes that are differently expressed in cancers compared
Enrique Baca-Garcia et al.
Progress in neuro-psychopharmacology & biological psychiatry, 28(2), 409-412 (2004-01-31)
This is the first case-control study exploring the association between suicide attempts and the polymorphic variations of the alpha 3 subunit gamma-aminobutyric acid (GABA) receptor gene (GABRA3) located in chromosome X. In a Spanish general hospital, 184 suicide attempters (127
Yan Liu et al.
World journal of gastroenterology, 14(47), 7175-7182 (2008-12-17)
To investigate the expression pattern of gamma-aminobutyric acid A (GABAA) receptors in hepatocellular carcinoma (HCC) and indicate the relationship among gamma-aminobutyric acid (GABA), gamma-aminobutyric acid A receptor alpha3 subunit (GABRA3) and HCC. HCC cell line Chang, HepG2, normal liver cell
Verena Henkel et al.
American journal of medical genetics. Part B, Neuropsychiatric genetics : the official publication of the International Society of Psychiatric Genetics, 126B(1), 82-87 (2004-03-30)
There is evidence that genes coding for the gamma aminobutyric acid (GABA) receptor may be involved in the etiology of affective disorders. Recently, an association between a CA-repeat in the GABRA 3 gene and bipolar disorder had been reported. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service