Skip to Content
Merck
All Photos(1)

Key Documents

H7278

Sigma-Aldrich

NG-Hydroxy-L-arginine acetate salt

Synonym(s):

NOHA acetate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14N4O3 · C2H4O2
CAS Number:
Molecular Weight:
250.25
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (TLC)

Quality Level

form

powder

mp

204 °C

solubility

acetic acid: 10 mg/mL, clear, colorless to faintly yellow

storage temp.

2-8°C

SMILES string

CC(O)=O.N[C@@H](CCCNC(=N)NO)C(O)=O

InChI

1S/C6H14N4O3.C2H4O2/c7-4(5(11)12)2-1-3-9-6(8)10-13;1-2(3)4/h4,13H,1-3,7H2,(H,11,12)(H3,8,9,10);1H3,(H,3,4)/t4-;/m0./s1

InChI key

VYMCYRPQICLHKC-WCCKRBBISA-N

Biochem/physiol Actions

Intermediate in the conversion of arginine to NO and citrulline by NO synthase.
NG-Hydroxy-L-arginine acts as a physiological inhibitor of arginase.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The inhibition of arginase by N?-hydroxy-L-arginine controls the growth of Leishmania inside macrophages
Iniesta V, et al.
The Journal of Experimental Medicine, 193(6), 777-784 (2001)
T Pera et al.
The Journal of pharmacology and experimental therapeutics, 349(2), 229-238 (2014-02-25)
Airway inflammation and remodeling are major features of chronic obstructive pulmonary disease (COPD), whereas pulmonary hypertension is a common comorbidity associated with a poor disease prognosis. Recent studies in animal models have indicated that increased arginase activity contributes to features
Alexander Bollenbach et al.
Analytical biochemistry, 577, 59-66 (2019-04-28)
l-Homoarginine (hArg) is biosynthesized from l-arginine (Arg) and l-lysine (Lys) by arginine:glycine amidinotransferase (AGAT). AGAT also catalyzes the formation of guanidinoacetate (GAA) from Arg and glycine (Gly). GAA is converted to creatine (N-methyl guanidinoacetate) by guanidinoacetate N-methyl-transferase (GAMT). Low circulating
K L Campos et al.
The Journal of biological chemistry, 270(4), 1721-1728 (1995-01-27)
The nitric oxide synthase-catalyzed conversion of L-arginine to L-citrulline and nitric oxide is known to be the sum of two partial reactions: oxygenation of arginine to N-hydroxyarginine, followed by oxygenation of N-hydroxyarginine to citrulline and nitric oxide. Whereas the conversion
P Klatt et al.
The Journal of biological chemistry, 268(20), 14781-14787 (1993-07-15)
Brain NO (nitric oxide) synthase contains FAD, FMN, heme, and tetrahydrobiopterin as prosthetic groups and represents a multi-functional oxidoreductase catalyzing oxidation of L-arginine to NO and L-citrulline, formation of H2O2, and reduction of cytochrome c. We show that substrate analogues

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service