Skip to Content
Merck
All Photos(2)

Documents

H4516

Sigma-Aldrich

Hydrocodone (+)-bitartrate salt

Synonym(s):

4,5-Epoxy-3-methoxy-17-methyl-5α-morphinan-6-one (2R,3R)-2,3-dihydroxybutanedioate (1:1)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H21NO3 · C4H6O6
CAS Number:
Molecular Weight:
449.45
EC Number:
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

drug control

USDEA Schedule II; Home Office Schedule 2; stupéfiant (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada; estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

solubility

H2O: 62 mg/mL
ethanol: soluble

application(s)

forensics and toxicology
veterinary

originator

Abbott

SMILES string

O[C@H]([C@@H](O)C(O)=O)C(O)=O.[H][C@]12CCC(=O)[C@]3([H])Oc4c(OC)ccc5C[C@H]1N(C)CC[C@@]23c45

InChI

1S/C18H21NO3.C4H6O6/c1-19-8-7-18-11-4-5-13(20)17(18)22-16-14(21-2)6-3-10(15(16)18)9-12(11)19;5-1(3(7)8)2(6)4(9)10/h3,6,11-12,17H,4-5,7-9H2,1-2H3;1-2,5-6H,(H,7,8)(H,9,10)/t11-,12+,17-,18-;1-,2-/m01/s1

InChI key

OJHZNMVJJKMFGX-BWCYBWMMSA-N

Biochem/physiol Actions

μ opioid receptor agonist; narcotic analgesic and antitussive.

Features and Benefits

This compound was developed by Abbott. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C J Kotzer et al.
The Journal of pharmacology and experimental therapeutics, 292(2), 803-809 (2000-01-20)
In this study, the activity of the delta-opioid receptor subtype-selective agonist, SB 227122, was investigated in a guinea pig model of citric acid-induced cough. Parenteral administration of selective agonists of the delta-opioid receptor (SB 227122), mu-opioid receptor (codeine and hydrocodone)
D M Tomkins et al.
The Journal of pharmacology and experimental therapeutics, 280(3), 1374-1382 (1997-03-01)
Humans that lack cytochrome P450 2D6 (CYP2D6) activity may have an altered risk of drug dependence or abuse because this enzyme is important in the metabolism of some drugs of abuse, including hydrocodone. In rats, hydrocodone conversion to hydromorphone is
Z R Chen et al.
Life sciences, 48(22), 2165-2171 (1991-01-01)
The binding affinity to the mu receptor of some opioids chemically related to morphine and some of their metabolites was examined in rat brain homogenates with 3H-DAMGO. The chemical group at position 6 of the molecule had little effect on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service