Skip to Content
Merck
All Photos(1)

Key Documents

H2395

Sigma-Aldrich

Hoagland′s No. 2 Basal Salt Mixture

powder, suitable for plant cell culture

Synonym(s):

Hoagland solution 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.72

form

powder

Quality Level

technique(s)

cell culture | plant: suitable

application(s)

agriculture

storage temp.

2-8°C

Application

Hoagland′s No. 2 Basal Salt Mixture has been used:
  • in the preparation of Hoagland′s solution for treating root tips
  • to grow maize (Zea mays L.) seeds
  • to grow rice seedlings and to treat M. sinensis (cv. Kosung) seedlings hydroponically

Hoagland′s No. 2 Basal Salt Mixture is a classic plant medium. Hoagland′s solution is a popular solution for a wide variety of plant species.

Formula variant

With the macro- and micronutrients as described by Hoagland and Arnon (1938).

Media Formulation

Quantity

Formulated to contain 1.6 grams of powder per liter of medium.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Ox. Sol. 3

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Renee S Arias et al.
BMC plant biology, 18(1), 170-170 (2018-08-17)
Aflatoxin contamination in peanut seeds is still a serious problem for the industry and human health. No stable aflatoxin resistant cultivars have yet been produced, and given the narrow genetic background of cultivated peanuts, wild species became an important source
Yunying Fang et al.
Global change biology, 24(7), 2775-2790 (2018-04-01)
Agronomic practices such as crop residue return and additional nutrient supply are recommended to increase soil organic carbon (SOC) in arable farmlands. However, changes in the priming effect (PE) on native SOC mineralization in response to integrated inputs of residue
The water-culture method for growing plants without soil.
Hoagland, D.R., and Arnon, D.I.
Univ. Calif. Coll. Agric. Exp. Sta. Circ., 347-353 (1938)
Dongling Mou et al.
Ecotoxicology and environmental safety, 74(4), 776-786 (2010-11-16)
The cuttings of grape (Vitis vinifera Linn.) were exposed to Hoagland's solution containing five different manganese (Mn) concentrations to investigate Mn toxicity and the possible detoxifying responses. Three genotypes (i.e. cultivars Combiner, Jingshou and Shuijing) were used in present study.
ZmCOI6. 1, a novel, alternatively spliced maize gene, whose transcript level changes under abiotic stress
Guerra-Peraza O, et al.
Plant Science, 176(6), 783-791 (2009)

Articles

Classical plant tissue culture media developed years ago by pioneers such as Murashige, Skoog, Gamborg, and others still play a vital role in plant tissue culture research today.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service