Skip to Content
Merck
All Photos(1)

Documents

H2387

SAFC

Hanks′ Balanced Salts

Modified, without calcium chloride, magnesium sulfate and sodium bicarbonate, powder, suitable for cell culture

Synonym(s):

HBSS

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75

form

powder

technique(s)

cell culture | mammalian: suitable

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Balanced salt solution (BSS) provides cells with water, bulk inorganic ions, and buffering system to maintain the normal cell metabolism and physiological range (pH 7.2-7.6), respectively. It also acts as an irrigating, diluting, and transporting agent and maintains the intracellular and extracellular osmotic balance. BSS also provides energy sources such as glucose for cell metabolism.

Application

Hanks′ Balanced Salt has been used:

  • to culture primary hippocampal neuron cells from rats
  • to collect the rat brains
  • to culture rat primary hepatocytes

Quantity

Formulated to contain 9.5 grams of powder per liter of medium.

Reconstitution

Supplement with 0.35 g/L sodium bicarbonate.

Other Notes

The pH is adjusted after addition of sodium bicarbonate.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abdelrahman I Rezk et al.
Scientific reports, 9(1), 117-117 (2019-01-16)
Magnesium (Mg) and its alloys have attracted much attention as a promising candidate for degradable implant applications however the rapid corrosion of magnesium inside the human body greatly limits its use as an implant material. Therefore, coating the alloy surface
Yu Bai et al.
Journal of materials science. Materials in medicine, 27(2), 31-31 (2015-12-26)
Au nanoparticles (AuNPs) are successfully assembled on TiO2 nanotube (TN) arrays through electrochemical deposition technology to improve the surface characteristics of TN arrays as an implant material. The loading amount of AuNPs can be controlled by adjusting the deposition time
Xiaojun Huang et al.
Progress in biophysics and molecular biology, 156, 3-13 (2020-08-08)
Cryo-electron microscopy (cryoEM) has become one of the most important approach for structural biology. However, barriers are still there for an increased successful rate, a better resolution and improved efficiency from sample preparation, data collection to image processing. CryoEM sample
Yu-Kyoung Kim et al.
Scientific reports, 8(1), 13264-13264 (2018-09-07)
A polymer coating as polycaprolactone (PCL) is applied to improve the initial corrosion resistance of biodegradable magnesium. In addition, plasma electrolytic oxidation (PEO) is performed to increase adhesion between the polymer and the metal. However, when a complex-shaped material such
Seo Yeon Lee et al.
Polymers, 12(2) (2020-02-20)
Herein, we describe precisely a covalent modification of pure magnesium (Mg) surface and its application to induce in vitro osteogenic differentiation. The new concept of a chemical bonding method is proposed for developing stable chemical bonds on the Mg surface

Protocols

Powdered media and salt mixtures are extremely hygroscopic and should be protected from atmospheric moisture.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service