Skip to Content
Merck
All Photos(3)

Key Documents

H1753

Sigma-Aldrich

Hydralazine hydrochloride

Synonym(s):

1-Hydrazinophthalazine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8N4 · HCl
CAS Number:
Molecular Weight:
196.64
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Assay

≥99% (TLC)

form

powder

mp

273 °C

solubility

water: 50 mg/mL

SMILES string

Cl[H].N\N=C1/N=NC=C2C=CC=CC12

InChI

1S/C8H8N4.ClH/c9-11-8-7-4-2-1-3-6(7)5-10-12-8;/h1-5,7H,9H2;1H/b11-8-;

InChI key

SECXUXOCDLQOBI-MKFZHGHUSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Hydralazine hydrochloride has been used:
  • as a vasodilator to study its effects on hypertension in T-cell frequencies in juvenile rats
  • as a semicarbazide-sensitive amine oxidase (SSAO) inhibitor to study its effects on myocardial ischemia-reperfusion (I/R) injury
  • as a vasodilator to study its effects on insulin secretion and glucose tolerance

Biochem/physiol Actions

Hydralazine hydrochloride has therapeutic effects against heart failure and high blood pressure.
Inhibits DNA methyltransferase and modulates epigenetic regulation of gene expression. Non-selective MAO-A/B inhibitor; antihypertensive; semicarbazide-sensitive amine oxidase inhibitor.

Features and Benefits

This compound is featured on the Dopamine and Norepinephrine Metabolism page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nadine S Sauter et al.
Diabetes, 64(4), 1273-1283 (2014-10-30)
Pathological activation of the renin-angiotensin system (RAS) is associated with the metabolic syndrome, and the new onset of type 2 diabetes can be delayed by RAS inhibition. In animal models of type 2 diabetes, inhibition of the RAS improves insulin
Ruijia Feng et al.
Gut microbes, 16(1), 2390164-2390164 (2024-08-18)
Non-alcoholic fatty liver disease (NAFLD) has emerged as a global health concern, lacking specific therapeutic strategies. Time-restricted feeding (TRF) regimen demonstrated beneficial effects in NAFLD; however, the underlying mechanisms remain unclear. In this study, we established a NAFLD mouse model
Edmond A Rogers et al.
Scientific reports, 12(1), 11838-11838 (2022-07-14)
While clinical observations have confirmed a link between the development of neurodegenerative diseases and traumatic brain injuries (TBI), there are currently no treatments available and the underlying mechanisms remain elusive. In response, we have developed an in vitro pendulum trauma
Philip C Burcham et al.
Molecular pharmacology, 82(5), 876-886 (2012-08-08)
Toxic carbonyls such as acrolein participate in many degenerative diseases. Although the nucleophilic vasodilatory drug hydralazine readily traps such species under "test-tube" conditions, whether these reactions adequately explain its efficacy in animal models of carbonyl-mediated disease is uncertain. We have
Mahesh P Kate et al.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism, 34(1), 81-86 (2013-09-21)
Blood pressure (BP) reduction after intracerebral hemorrhage (ICH) is controversial, because of concerns that this may cause critical reductions in perihematoma perfusion and thereby precipitate tissue damage. We tested the hypothesis that BP reduction reduces perihematoma tissue oxygenation.Acute ICH patients

Articles

Cancer research has revealed that the classical model of carcinogenesis, a three step process consisting of initiation, promotion, and progression, is not complete.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service