Skip to Content
Merck
All Photos(3)

Key Documents

H0513

Sigma-Aldrich

Heptakis(2,6-di-O-methyl)-β-cyclodextrin

Synonym(s):

DM-β-CD, DM‐β‐CyD, heptakis-(2,6-O-Dimethyl)-β-cyclodextrin, 2,6-Di-O-methyl-β-cyclodextrin, Dimethyl β-cyclodextrin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C56H98O35
CAS Number:
Molecular Weight:
1331.36
Beilstein:
1679144
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

biological source

synthetic

Quality Level

form

powder

SMILES string

COC[C@H]1O[C@@H]2O[C@H]3[C@H](O)[C@@H](OC)[C@H](O[C@@H]3COC)O[C@H]4[C@H](O)[C@@H](OC)[C@H](O[C@@H]4COC)O[C@H]5[C@H](O)[C@@H](OC)[C@H](O[C@@H]5COC)O[C@H]6[C@H](O)[C@@H](OC)[C@H](O[C@@H]6COC)O[C@H]7[C@H](O)[C@@H](OC)[C@H](O[C@@H]7COC)O[C@H]8[C@H](O)[C@@H](OC)[C@H](O[C@@H]8COC)O[C@H]1[C@H](O)[C@H]2OC

InChI

1S/C56H98O35/c1-64-15-22-36-29(57)43(71-8)50(78-22)86-37-23(16-65-2)80-52(45(73-10)30(37)58)88-39-25(18-67-4)82-54(47(75-12)32(39)60)90-41-27(20-69-6)84-56(49(77-14)34(41)62)91-42-28(21-70-7)83-55(48(76-13)35(42)63)89-40-26(19-68-5)81-53(46(74-11)33(40)61)87-38-24(17-66-3)79-51(85-36)44(72-9)31(38)59/h22-63H,15-21H2,1-14H3/t22-,23-,24-,25-,26-,27-,28-,29+,30+,31+,32+,33+,34+,35+,36-,37-,38-,39-,40-,41-,42-,43-,44-,45-,46-,47-,48-,49-,50-,51-,52-,53-,54-,55-,56-/m1/s1

InChI key

QGKBSGBYSPTPKJ-UZMKXNTCSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Heptakis(2,6-di-O-methyl)-β-cyclodextrin is a hydrophilic derivative of cyclodextrin that possesses the ability to effectively dissolve lipophilic drugs like steroid hormones, vitamins A, E, and K, etc. It can be utilized to increase the dissolution and bioavailability of weakly water-soluble pharmaceutical compounds.

Application

Heptakis(2,6-di-O-methyl)-β-cyclodextrin may be used:
  • To measure the equilibrium constant of its complexes with various enantiomeric pairs of chiral nitroxides by electron paramagnetic resonance (EPR) spectroscopy.
  • To study the crystal structure of its complex with n-butyl acrylate and isobornyl acrylate using X-ray diffraction (XRD).
  • As a chiral selector to resolve amphetamine related drugs by capillary zone electrophoresis.

Reactant involved in:
  • Physicochemical studies of inclusion complexes
  • Probing diffusion and single molecule interactions with reconstituted membrane proteins
  • Interactions with micelles causing micellar rupture
  • Studying cellular transport, used as a cell penetration enhancer
  • Physicochemical and biopharmaceutical improvement of drugs particularly with solubilization and stability
  • Interactions with cholesterol

Quality

Impurities are over-methylated homologs, primarily hexakis(2,6-di-O-methyl)-(2,3,6-tri-O-methyl)-β-cyclodextrin.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Paola Franchi et al.
Journal of the American Chemical Society, 126(13), 4343-4354 (2004-04-01)
EPR spectroscopy has been employed to investigate the formation of complexes between heptakis-(2,6-O-dimethyl)-beta-cyclodextrin (DM-beta-CD) and different enantiomeric pairs of chiral nitroxides of general structure PhCH2NO.CH(R)R'. Accurate equilibrium measurements of the concentrations of free and included radicals afforded the binding constant
F Karamustafa et al.
Die Pharmazie, 64(2), 98-103 (2009-03-27)
The transport of alendronate through Caco-2 monolayers in the absence and presence of absorption enhancers (sodium taurocholate-STC and dimethyl-beta-cyclodextrin-DM-beta-CD) was studied. The viability of Caco-2 cells was determined by MTT assay. The effects of the experiment period and serum existence
Anita Zamboni et al.
BMC genomics, 10, 363-363 (2009-08-08)
In response to pathogen attack, grapevine synthesizes phytoalexins belonging to the family of stilbenes. Grapevine cell cultures represent a good model system for studying the basic mechanisms of plant response to biotic and abiotic elicitors. Among these, modified beta-cyclodextrins seem
Enantiomeric separation of amphetamine related drugs by capillary zone electrophoresis using native and derivatized beta-cyclodextrin as chiral additives
Cladrowa-Runge S, et al.
Journal of Chromatography A, 710(2), 339-345 (1995)
Fei Liu et al.
Chemical & pharmaceutical bulletin, 60(12), 1479-1486 (2012-12-05)
Thienorphine hydrochloride (ThH) is a highly insoluble and readily metabolized partial-opioid agonist. It is used for the treatment of pain and heroin addiction. This study aimed to formulate and evaluate sublingual delivery systems containing ThH. Dimethyl-β-cyclodextrin (DM-β-CD) can enhance the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service