GE17-1440-03
Ficoll® Paque Plus
Cytiva 17-1440-03, pack of 6 × 500 mL
Synonym(s):
Ficoll density gradient medium, cell culture media
About This Item
Recommended Products
form
liquid
packaging
pack of 6 × 500 mL
manufacturer/tradename
Cytiva 17-1440-03
color
Colorless to slightly yellow
pH range
5.5-7.5
General description
Ficoll-Paque PLUS is a ready to use, sterile medium for isolation of lymphocytes in high yield from peripheral blood using a simple and rapid centrifugation procedure. It maintains the viability and a representative distribution of B and T lymphocytes.
Application
Features and Benefits
- Ready to use sterile medium for isolation of human lymphocytes in high yield from peripheral blood.
- Maintains viability and representative distribution of B and T lymphocytes.
- Low levels of endotoxin (< 0.12 EU/mL).
Storage and Stability
Analysis Note
Legal Information
Storage Class Code
12 - Non Combustible Liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service