Skip to Content
Merck
All Photos(1)

Key Documents

G7652

Sigma-Aldrich

Anti-Mouse IgG (whole molecule)–Gold antibody produced in goat

affinity isolated antibody, aqueous glycerol suspension, 10 nm (colloidal gold)

Synonym(s):

Goat anti-mouse IgG

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

gold conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

aqueous glycerol suspension

particle size

10 nm (colloidal gold)

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Binding activity is determined using a modification of the dot blot assay of Brada and Roth. In this assay a 1 mg/mL solution of mouse IgG is serially diluted in phosphate buffered saline, 1 mL of each dilution is then applied to nitrocellulose paper. Binding activity is defined as the minimum amount of mouse IgG detectable as a visible pink-red spot after a 1 hour incubation with the gold conjugate diluted to an A520 of0.25. Relative order of singlets, doublets, triplets or greater: Singlets>Doublets>Triplets. Clustering data determined by floating the electron microscopy (EM) nickel grid on conjugate diluted 1:100 in Tris buffer, pH 8.0, containing 1% BSA, for 30-60 minutes. The grid is evaluated by EM after drying. One hundred particles are scanned to determine clustering.
Immunoglobulin G (IgG) is a glycoprotein antibody that regulates immune responses such as phagocytosis and is also involved in the development of autoimmune diseases . Mouse IgGs have four distinct isotypes, namely, IgG1, IgG2a, IgG2b, and IgG3. IgG1 regulates complement fixation in mice . Anti-Mouse IgG (whole molecule)-Gold antibody is specific for mouse IgG.
Mouse IgG is a plasma B cell derived antibody isotype defined by its heavy chain. IgG is the most abundant antibody isotype found in mouse serum. IgG crosses the placental barrier, is a complement activator and binds to the Fc-receptors on phagocytic cells. The level of IgG may vary with the status of disease or infection.
Immunogold labeling is a technique whereing antibodies, especially secondary antibodies, are labeled with colloidal gold. These gold conjugated antibodies generate electron dense particles that are visable using electron microscopy.

Immunogen

Mouse IgG purified from normal mouse serum

Application

Anti-Mouse IgG (whole molecule)-Gold antibody is suitable for use in inductively coupled plasma mass spectrometry (ICPMS)-Linked DNA Assay ((1:100). The product can also be used for immunoelectron microscopy (1:20).
Goat polyclonal anti-mouse IgG (whole molecule)–Gold antibody may be used as a primary or secondary antibody that enables the labeling of mouse IgG for detection by electron microscopy.

Physical form

Colloidal suspension in Tris buffered saline, pH 8.2, with 30% glycerol (v/v), 1% bovine serum albumin (w/v), and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arben Merkoçi et al.
Analytical chemistry, 77(19), 6500-6503 (2005-10-01)
Gold nanoparticles modified with anti-mouse IgG have been used to trace oligonucleotides carrying a c-myc peptide. Two strategies, a dot-blot format as well as inductively coupled plasma mass spectrometry (ICPMS) have been used to detect the nanoparticle tracer. For both
Yun Zhang et al.
Molecular therapy : the journal of the American Society of Gene Therapy, 6(1), 67-72 (2002-07-04)
Therapeutic genes are delivered to the nuclear compartment of cancer cells following intravenous administration with a non-immunogenic "artificial virus" gene delivery system that uses receptor-specific monoclonal antibodies (MAb) to navigate the biological barriers between the blood and the nucleus of
Daryl E Enstone et al.
Scientific reports, 5, 15074-15074 (2015-10-13)
Hydrophobic protein from soybean (HPS) is present in soybean dust and is an allergen (Gly m 1) that causes asthma in allergic individuals. Past studies have shown that HPS occurs on the seed surface. To determine the microscopic localization of
Xiaoliang Shi et al.
Journal of integrative plant biology, 60(2), 112-129 (2017-12-07)
Since its domestication from wild rice thousands of years ago, rice has been cultivated largely through transplantation. During transplantation from the nursery to the paddy field, rice seedlings experience transplantation shock which affects their physiology and production. However, the mechanisms
Jin-Yong Noh et al.
PloS one, 11(9), e0162946-e0162946 (2016-09-15)
Highly pathogenic avian influenza (HPAI) and Newcastle disease (ND) are considered as the most devastating poultry infections, owing to their worldwide distribution and economical threat. Vaccines have been widely used to control these diseases in the poultry industry in endemic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service