Skip to Content
Merck
All Photos(8)

Key Documents

G7141

Sigma-Aldrich

Glucose Oxidase from Aspergillus niger

greener alternative

Type X-S, lyophilized powder, 100,000-250,000 units/g solid (without added oxygen)

Synonym(s):

β-D-Glucose:oxygen 1-oxidoreductase, G.Od., GOx

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54
Pricing and availability is not currently available.

type

Type X-S

Quality Level

form

lyophilized powder

specific activity

100,000-250,000 units/g solid (without added oxygen)

mol wt

160 kDa

composition

Protein, ≥65%

greener alternative product characteristics

Waste Prevention
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

application(s)

diagnostic assay manufacturing

foreign activity

Catalase ≤5 units/mg protein

greener alternative category

storage temp.

−20°C

InChI

1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5-,6-/m1/s1

InChI key

WQZGKKKJIJFFOK-VFUOTHLCSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Mass of glucose oxidase (GOX), a flavoprotein ranges from approximately 130 to 175 kDa.[1] Some fungi and insects are capable of producing GOX.[2]
Molecular Weight: 160 kDa (gel filtration)
pI: 4.2
Extinction coefficient: E1% = 16.7 (280 nm)

Glucose oxidase from Aspergillus niger is a dimer consisting of 2 equal subunits with a molecular mass of 80 kDa each. Each subunit contains one flavin adenine dinulceotide moiety and one iron. The enzyme is a glycoprotein containing ~16% neutral sugar and 2% amino sugars. The enzyme also contains 3 cysteine residues and 8 potential sites for N-linked glycosylation.

Glucose oxidase is capable of oxidizing D-aldohexoses, monodeoxy-D-glucoses, and methyl-D-glucoses at varying rates.

The pH optimum for glucose oxidase is 5.5, while it has a broad activity range of pH 4-7. Glucose oxidase is specific for β-D-glucose with a KM of 33-110 mM.

Glucose oxidase does not require any activators, but it is inhibited by Ag+, Hg2+, Cu2+, phenylmercuric acetate, and p-chloromercuribenzoate. It is not inhibited by the nonmetallic SH reagents: N-ethylmaleimide, iodoacetate, and iodoacetamide.

Glucose oxidase can be utilized in the enzymatic determination of D-glucose in solution. As glucose oxidase oxidizes β-D-glucose to D-gluconolactate and hydrogen peroxide, horseradish peroxidase is often used as the coupling enzyme for glucose determination. Although glucose oxidase is specific for β-D-glucose, solutions of D-glucose can be quantified as α-D-glucose will mutorotate to β-D-glucose as the β-D-glucose is consumed by the enzymatic reaction.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency and waste prevention when used in fuel cell research. For more information see the article in biofiles.

Application

Glucose Oxidase from Aspergillus niger has been used:
  • in the (glucose oxidase) GO reagent to measure the glucose content by the glucose oxidase (GO) method[3]
  • to activate the human renal carcinoma cell line for constructing the oxidative stress model[4]
  • to study its influence in the paste on the analytical performance of the bioelectrode[5]

Glucose oxidase is widely used in the food and pharmaceutical industries as well as a major component of glucose biosensors.

Biochem/physiol Actions

Glucose oxidase (GOX) oxidizes D-glucose to D-gluconolactone and hydrogen peroxide. Hydrogen peroxide, the catalytic product of GOX, serves as an anti-bacterial and anti-fungal agent.[2] It is safe and can be used in several industrial applications like baking, dry egg powder production, wine production, gluconic acid production, etc. It possesses electrochemical activity, which makes it extremely important in glucose sensors and in fuel cell applications.[2]
Glucose oxidase catalyses the oxidation of β-d-glucose to d-glucono-β-lactone and hydrogen peroxide, with molecular oxygen as an electron acceptor.

Quality

May contain traces of amylase, maltase, glycogenase, invertase, and galactose oxidase.

Unit Definition

One unit will oxidize 1.0 μmole of β-D-glucose to D-gluconolactone and H2O2 per min at pH 5.1 at 35 °C, equivalent to an O2 uptake of 22.4 μl per min. If the reaction mixture is saturated with oxygen, the activity may increase by up to 100%.

Analysis Note

Protein determined by biuret

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Interspecies Interactions Determine the Impact of the Gut Microbiota on Nutrient Allocation in Drosophila melanogaster
Newell PD, et al.
Applied and Environmental Microbiology, 80(2), 788-796 (2014)
Glucose biosensors based on the immobilization of copper oxide and glucose oxidase within a carbon paste matrix
Luque GL, et al.
Talanta, 66(2), 467-471 (2005)
Raluca-Elena Munteanu et al.
Scientific reports, 9(1), 15196-15196 (2019-10-28)
If the analyte does not only change the electrochemical but also the optical properties of the electrode/solution interface, the spatial resolution of an electrochemical sensor can be substantially enhanced by combining the electrochemical sensor with optical microscopy. In order to
Glucose oxidase-An overview
Biotechnology Advances, 27(4), 489-501 (2009)
Ewa Szczęsna et al.
European journal of cell biology, 95(12), 521-530 (2016-09-10)
End-binding proteins are capable of tracking the plus-ends of growing microtubules (MTs). The motor protein Ncd, a member of the kinesin-14 family, interacts with EB1 protein and becomes a non-autonomous tip-tracker. Here, we attempted to find out whether at least

Protocols

Glucose oxidase activity measured via continuous spectrophotometric assay at 500 nm, indicating glucose oxidation rate.

Glucose oxidase activity measured via continuous spectrophotometric assay at 500 nm, indicating glucose oxidation rate.

Glucose oxidase activity measured via continuous spectrophotometric assay at 500 nm, indicating glucose oxidation rate.

Glucose oxidase activity measured via continuous spectrophotometric assay at 500 nm, indicating glucose oxidation rate.

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service