Skip to Content
Merck
All Photos(1)

Key Documents

G5001

Sigma-Aldrich

DL-Glyceraldehyde

≥90% (GC)

Synonym(s):

α,β-Dihydroxypropionaldehyde, 2,3-Dihydroxypropanal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H6O3
CAS Number:
Molecular Weight:
90.08
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic

Assay

≥90% (GC)

form

powder

color

white to off-white

mp

145  °C ((293 °F ))

solubility

water: 50 mg/mL, clear to slightly hazy, colorless to faintly yellow

storage temp.

room temp

SMILES string

[H]C(=O)C(O)CO

InChI

1S/C3H6O3/c4-1-3(6)2-5/h1,3,5-6H,2H2

InChI key

MNQZXJOMYWMBOU-UHFFFAOYSA-N

General description

Glyceraldehyde is a simple monosaccharide. Based on the number of carbon atoms and the type of carbonyl group present, glyceraldehyde belongs to subgroup triose. It is a colourless and sweet compound.

Application

DL-Glyceraldehyde has been used:
  • as modifying reagent in the preparation of crystallization solution
  • as a substrate to measure aldose reductase activity
  • in the preparation of d/l-glyceraldehyde stock to determine the specific activity of GAPDH (glyceraldehyde 3-phosphate dehydrogenase)

Biochem/physiol Actions

Glyceraldehyde serves as an efficient cross-linking agent and is considered non-toxic. It is an intermediate of a number of metabolic such as glycolysis and pentose phosphate pathway.

Other Notes

DL-Glyceraldehyde is a substrate for the enzyme aldose reductase.
To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raman and infrared spectroscopy of carbohydrates: a review
Wiercigroch E, et al.
Spectrochimica Acta. Part A, Molecular and Biomolecular Spectroscopy, 185(5), 317-335 (2017)
Gemma Sangüesa et al.
European journal of nutrition, 58(3), 1283-1297 (2018-03-09)
Sugar-sweetened beverage intake is a risk factor for insulin resistance, dyslipidemia, fatty liver, and steatohepatitis (NASH). Sub-chronic supplementation of liquid fructose, but not glucose, in female rats increases liver and plasma triglycerides without inflammation. We hypothesized that chronic supplementation of
In-Geun Ryoo et al.
Redox biology, 17, 246-258 (2018-05-08)
Cluster of differentiation 44 (CD44) is the most common cancer stem cell (CSC) marker and high CD44 expression has been associated with anticancer drug resistance, tumor recurrence, and metastasis. In this study, we aimed to investigate the molecular mechanism by
Jiarui Sun et al.
Research in veterinary science, 109, 135-141 (2016-11-29)
Several studies have found that melamine causes damage to the testes, epididymis and sperm. However, few studies have investigated the effect of melamine on the synthesis of testosterone, which plays an import role in testicular development and spermatogenesis. In present
Stabilization of scleral collagen by glycerol aldehyde cross-linking
N.A.Danilov, et al.
Biochimica et Biophysica Acta, 1780(5), 764-772 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service