Skip to Content
Merck
All Photos(1)

Key Documents

G3018

Sigma-Aldrich

Asialoganglioside GM1 from bovine brain

~98%, lyophilized powder

Synonym(s):

Ceramide tetrahexoside, Ganglioside GM1, asialo, Gangliotetraosyl ceramide

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

~98%

Quality Level

form

lyophilized powder

storage temp.

−20°C

InChI

1S/C62H114N2O23/c1-4-6-8-10-12-14-16-18-19-21-23-25-27-29-31-33-46(71)64-40(41(70)32-30-28-26-24-22-20-17-15-13-11-9-7-5-2)38-80-60-54(78)51(75)57(44(36-67)83-60)86-62-55(79)52(76)56(45(37-68)84-62)85-59-47(63-39(3)69)58(49(73)43(35-66)81-59)87-61-53(77)50(74)48(72)42(34-65)82-61/h30,32,40-45,47-62,65-68,70,72-79H,4-29,31,33-38H2,1-3H3,(H,63,69)(H,64,71)/b32-30+/t40?,41?,42-,43-,44-,45-,47-,48+,49+,50+,51-,52-,53-,54-,55-,56+,57-,58-,59+,60?,61+,62+/m1/s1

InChI key

VELGMVLNORPMAO-JTFNWEOFSA-N

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Cer-Glc-Gal-GalNAc-Gal

General description

Gangliosides are major constituents of neuronal cell membranes and endoplasmic reticulum; contain a sialylated polysaccharide chain linked to ceramide through a β-glycosidic linkage; for classification of gangliosides see Svennerholm, L., et al. (eds.), Structure and Function of Gangliosides, New York, Plenum, 1980.

Biochem/physiol Actions

Removing the sialic acid from monosialoganglioside GM1 inhibits its ability to bind cholera toxin. Asialoganglioside GM1 does not block the binding of cholera toxin to cellular GM1.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julia Pinkhasov et al.
Cancer immunology, immunotherapy : CII, 59(12), 1801-1811 (2010-09-09)
Since epithelial mucin 1 (MUC1) is associated with several adenocarcinomas at the mucosal sites, it is pertinent to test the efficacy of a mucosally targeted vaccine formulation. The B subunit of the Vibrio cholerae cholera toxin (CTB) has great potential
A Abulrob et al.
Journal of microscopy, 232(2), 225-234 (2008-11-20)
The localization of asialo-GM1 in ordered membrane raft domains in HeLa cells has been examined using a combination of membrane fractionation and fluorescence imaging. The glycolipid is enriched in Triton X-100 insoluble membrane fractions that contain high concentrations of cholesterol
Hideto Nishikado et al.
Journal of immunology (Baltimore, Md. : 1950), 186(10), 5766-5771 (2011-04-15)
NK cells are innate immune lymphocytes and play a key role in both innate and adaptive immunity. Their pivotal functions in vivo have been illustrated in mice by means of their ablation with NK cell-depleting Abs, particularly anti-asialo GM1 (ASGM1).
Adam W Mailloux et al.
International journal of cancer, 127(11), 2598-2611 (2010-03-04)
Tumor-induced immune suppression involves the accumulation of suppressive infiltrates in the tumor microenvironment such as regulatory T-cells (Tregs). Previous studies demonstrated that NK-dependant increases in CCL22 secretion selectively recruit Tregs toward murine lungs bearing Lewis Lung Carcinoma (LLC). To extend
Pawel R Kiela et al.
Gastroenterology, 137(3), 965-975 (2009-05-20)
NHE3 is a target of inhibition by proinflammatory cytokines and pathogenic bacteria, an event contributing to diarrhea in infectious and idiopathic colitis. In mice, NHE3 deficiency leads to mild diarrhea, increased intestinal expression of interferon (IFN)-gamma, and distal colitis, suggesting

Articles

Understand sialic acid structure, function, signaling, and modifications. Easily find products for sialic acid research.

Understand sialic acid structure, function, signaling, and modifications. Easily find products for sialic acid research.

Understand sialic acid structure, function, signaling, and modifications. Easily find products for sialic acid research.

Understand sialic acid structure, function, signaling, and modifications. Easily find products for sialic acid research.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service