Skip to Content
Merck
All Photos(1)

Key Documents

G1041

Sigma-Aldrich

EZBlue Gel Staining Reagent

Synonym(s):

protein gel stain, protein stain

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105300
NACRES:
NA.32

technique(s)

protein staining: suitable

storage temp.

2-8°C

General description

Convenient, sensitive, and safe, EZBlue Coomassie Brilliant Blue G-250 colloidal protein stain improves protein electrophoresis results while significantly reducing staining time. Conveniently packaged, EZBlue requires no messy weigh-ups or additions of methanol or acid. As a colloidal stain, it reacts only with proteins, not the gel itself. Background staining is reduced, so protein bands can be visualized almost immediately. No destaining step is required, although a water wash may intensify bands and clarify the background. Most impressively, EZBlue is extremely sensitive, detecting as little as 5 ng of protein.

Application

EZBlue Gel Staining Reagent has been used as a staining reagent in sodium dodecyl sulfate polyacrylamide gel electrophoresis (SDS-PAGE).

Features and Benefits

  • Premixed solution eliminates the time and effort required to prepare the stain
  • Increased sensitivity ensures that low abundance proteins can be detected (as little as 5 ng)
  • Rapid reaction significantly reduces the amount of time required to stain and rinse
  • No solvent waste so you save time and money by eliminating hazardous material disposal

Other Notes

A ready-to-use Brilliant Blue G-250 based protein stain for "one step" ultrasensitive detection on polyacrylamide gels and PVDF membranes.

Legal Information

EZBlue is a trademark of Sigma-Aldrich Co. LLC
SIGMA-ALDRICH is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardCorrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - STOT SE 2

Target Organs

Eyes,Central nervous system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

AMPA receptor antibodies in limbic encephalitis alter synaptic receptor location.
Lai M, et al.
Annals of Neurology, 65, 424-434 (2009)
Praveen Maurye et al.
Electrophoresis, 38(16), 2060-2068 (2017-04-27)
PAGE is the most widely used technique for the separation and biochemical analysis of biomolecules. The ever growing field of proteomics and genomics necessitates the analysis of many proteins and nucleic acid samples to understand further about the structure and
Rakhi Panda et al.
Journal of agricultural and food chemistry, 63(43), 9629-9639 (2015-10-09)
Many soybean protein products are processed by enzymatic hydrolysis to attain desirable functional food properties or in some cases to reduce allergenicity. However, few studies have investigated the effects of enzymatic hydrolysis on the allergenicity of soybean products. In this
Antibodies to the GABA(B) receptor in limbic encephalitis with seizures: case series and characterisation of the antigen.
Lancaster E, et al.
Lancet Neurology, 9, 67-76 (2010)
Nathan L Marsteller et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 141, 111398-111398 (2020-05-22)
Currently no validated animal model is predictive of human responses in ranking purified dietary proteins in the prevalence or potency of food allergy in humans. Since the gastrointestinal microbiota is thought to influence oral tolerance, we hypothesize that a germ-free

Articles

Cell lysis challenges and protein extraction variables for Western blot sensitivity and reproducibility.

Cell lysis challenges and protein extraction variables for Western blot sensitivity and reproducibility.

Cell lysis challenges and protein extraction variables for Western blot sensitivity and reproducibility.

Cell lysis challenges and protein extraction variables for Western blot sensitivity and reproducibility.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service