Skip to Content
Merck
All Photos(1)

Key Documents

G0290

Sigma-Aldrich

Anti-GATA1 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-ERYF1, Anti-Erythroid transcription factor 1, Anti-Globin transcription factor 1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 45 kDa

species reactivity

human

packaging

antibody small pack of 25 μL

technique(s)

microarray: suitable
western blot: 1:200 using a whole extract from a human chronic myelogenous leukemia cell line, K562

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GATA1(2623)

Looking for similar products? Visit Product Comparison Guide

General description

GATA is an important cis-element factor in the haematopoietic lineage belongs to GATA-binding protein family. It was first identified in the analysis of globin gene promoters and enhancers. There are different types of GATA proteins: GATA-1, GATA-2, GATA-3. It is expressed in erythroid, mast, megakaryocytic lineages, multipotential progenitor cells and lines.
GATA-1 (ERYF1, GF-1, NF-1) is a Cys2/Cys2 zinc finger DNA binding protein that is expressed primarily in erythroid, megakaryocytic, mast cells and eosinophilic cells.

Immunogen

synthetic peptide corresponding to the C-terminal region of human GATA-1 (amino acids 394-413).

Application

Anti-GATA1 antibody produced in rabbit has been used in gel mobility shift assay and DNase I footprinting. It has also been used in confocal microscopy analysis.

Biochem/physiol Actions

GATA is a lineage-restricted transcription factor which plays a pivotal role in erythroid differentiation. It possesses potential binding sites in the regulatory sequences of all erythroid genes. GATA is expressed at higher levels during erythroid maturation. It is essential for terminal differentiation of erythrocytes and megakaryocytes and associated in vivo with the acetyl transferase p300/CBP. Mutation in GATA1 causes thrombocytopenia and abnormal platelet function.
GATA-binding factor 1 (GATA-1) is involved in blocking apoptosis of precursor cells and in controlling the balance between proliferation and cell cycle arrest.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CREB-binding protein cooperates with transcription factor GATA-1 and is required for erythroid differentiation
Blobel GA, et al.
Proceedings of the National Academy of Sciences of the USA, 95(5), 2061-2066 (1998)
M E Cullen et al.
The Journal of biological chemistry, 272(4), 2464-2469 (1997-01-24)
We have set out to test a model for tissue-specific gene expression that relies on the early replication of expressed genes to sequester limiting activating transcription factors. Using an erythroid cell line, we have tested the changes in the DNA
GATA-binding transcription factors in hematopoietic cells.
S H Orkin
Blood, 80(3), 575-581 (1992-08-01)
Erythropoietin and IGF-1 signaling synchronize cell proliferation and maturation during erythropoiesis
Kadri Z, et al.
Genes & Development, 29(24), 2603-2616 (2015)
Sascha C Hughan et al.
Blood, 105(11), 4369-4376 (2005-02-11)
Defects in the X-linked DNA-binding megakaryocyte transcription factor GATA1 cause thrombocytopenia and abnormal platelet function. However, detailed studies of GATA1 function in platelet activation are lacking. Here, we studied platelets from GATA1-deficient mice and from a male patient (S14) with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service