Skip to Content
Merck
All Photos(1)

Key Documents

F8924

Sigma-Aldrich

Fibroblast Growth Factor-10 human

>97% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

Synonym(s):

FGF-10

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

Quality Level

recombinant

expressed in E. coli

Assay

>97% (SDS-PAGE)

form

lyophilized powder

potency

28 ng/mL

mol wt

19 kDa by SDS-PAGE

packaging

pkg of 25 μg

storage condition

avoid repeated freeze/thaw cycles

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... FGF10(2255)

General description

FGF10 (fibroblast growth factor 10) is a member of the FGF protein family, which is a mitogen of epithelial origin and has a paracrine activity. It is produced by mesenchymal lineage cells, and acts as a ligand for FGFR, most notably FGFR2b, which is expressed by epithelial cells.

Application

Fibroblast Growth Factor-10 human is suitable for culturing of HUVEC cell line (human umbilical vein endothelial cells) to determine the anti-angiogenic properties of sFLT-1 (soluble fms-related tyrosine kinase-1).

Biochem/physiol Actions

FGF10 (fibroblast growth factor 10) is involved in cell growth, morphogenesis and embryogenesis. It plays an essential role in initiating the formation of limb bud, lung morphogenesis and brain formation during embryogenesis. It acts as a mitogen, where it facilitates the regeneration of epithelia post-bladder urothelial injury. Thus, it might have future applications during bladder transplantation. Studies in Japanese population show that polymosphisms- rs339501, rs12517396, and rs10462070 are linked with severe myopia, and might act as susceptibility factors for the same. In lung cancer cells, FGFR4 reduces the stem cell/self-renewal property of cancer cells, and makes them more susceptible to chemotherapy.

Physical form

Lyophilized from a 0.2 μm filtered solution in 20 mM MOPS, 50 mM sodium sulfate, 1 mM DTT, 0.5 mM EDTA, pH 7.2, containing 50 μg bovine serum albumin per 1 μg cytokine.

Analysis Note

The biological activity is measured by its ability to stimulate the proliferation of a monkey epithelial cell line 4MBr-5.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Poopak Farnia et al.
International journal of clinical and experimental medicine, 8(1), 1526-1533 (2015-03-19)
The soluble vascular endothelial growth factor receptor-1 (VEGFR1) or sFLT-1 has important role in antiangiogenesis. In this study, the increase expression and production of sFLT-1 fragment by newly designed ChPL-NPs nanoparticles (chitosan-protein lipid) using Chinese hamster ovary cell line (CHO)
Seyung S Chung et al.
In vitro cellular & developmental biology. Animal, 49(10), 746-751 (2013-08-21)
Fibroblast growth factor 10 (FGF10) is required for embryonic epidermal morphogenesis including brain development, lung morphogenesis, and initiation of limb bud formation. In this study, we investigated the role of FGF10 as a lead induction factor for stem cell differentiation
Masao Yoshida et al.
Molecular vision, 19, 2321-2329 (2013-11-23)
The fibroblast growth factor 10 (FGF10) gene polymorphism rs339501 was previously reported to be associated with high myopia in a Chinese population. In the present study, we investigated whether FGF10 polymorphisms are associated with extreme myopia in a Japanese population
H Ohuchi et al.
Development (Cambridge, England), 124(11), 2235-2244 (1997-06-01)
Vertebrate limb formation has been known to be initiated by a factor(s) secreted from the lateral plate mesoderm. In this report, we provide evidence that a member of the fibroblast growth factor (FGF) family, FGF10, emanates from the prospective limb
S Tagashira et al.
Gene, 197(1-2), 399-404 (1997-10-23)
We cloned the mouse homolog of FGF10, which was recently reported as a new member of the FGF family. The predicted molecular mass of this molecule is 23.6 kDa, and both nucleotide and amino acid sequences show high degrees of

Articles

Role of growth factors in stem cell differentiation and various growth factors for your research at sigmaaldrich.com

Role of growth factors in stem cell differentiation and various growth factors for your research at sigmaaldrich.com

Role of growth factors in stem cell differentiation and various growth factors for your research at sigmaaldrich.com

Role of growth factors in stem cell differentiation and various growth factors for your research at sigmaaldrich.com

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service