Skip to Content
Merck
All Photos(1)

Key Documents

F8182

Sigma-Aldrich

Faropenem sodium hydrate

≥98% (HPLC)

Synonym(s):

(5R,6S,8R,2′R)-2-(2′-tetrahydrofuryl)-6-hydroxyethylpenem-3-carboxylate sodium salt, ALP 201, Farom, Fropenem, Furopenem, SUN 5555, SY 5555, Wy 49605

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14NNaO5S · xH2O
CAS Number:
Molecular Weight:
307.30 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D +120 to +130°, c = 1.0 in water

storage condition

desiccated

color

white to light brown

solubility

H2O: ≥20 mg/mL

originator

Daiichi-Sankyo

storage temp.

−20°C

SMILES string

O.[Na+].C[C@@H](O)C1C2SC(C3CCCO3)=C(N2C1=O)C([O-])=O

InChI

1S/C12H15NO5S.Na.H2O/c1-5(14)7-10(15)13-8(12(16)17)9(19-11(7)13)6-3-2-4-18-6;;/h5-7,11,14H,2-4H2,1H3,(H,16,17);;1H2/q;+1;/p-1/t5-,6-,7+,11-;;/m1../s1

InChI key

FHSVCMPZCIOKGW-VIDQLUEFSA-M

Related Categories

General description

Faropenem sodium hydrate belongs to the penem group of antibiotics prescribed for oral usage. Enterobacteriaceae bacterial infections with cephalosporin resistance are susceptible to faropenem. Faropenem could be an effective antibiotic to treat urinary tract infections caused by extended-spectrum beta-lactamases (ESBL) producing bacteria.

Biochem/physiol Actions

Broad spectrum beta-lactam antibiotic belonging to the penem group; β-lactamase stable.
Faropenem sodium is an ultra-broad spectrum, β-lactamase resistant, β-lactam antibiotic active against both Gram-positive and Gram-negative bacteria.

Features and Benefits

This compound was developed by Daiichi-Sankyo. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Faropenem, a new oral penem: antibacterial activity against selected anaerobic and fastidious periodontal isolates.
Milazzo I, et al.
The Journal of Antimicrobial Chemotherapy, 51(3), 721-725 (2003)
S K Spangler et al.
Antimicrobial agents and chemotherapy, 38(11), 2599-2604 (1994-11-01)
The National Committee for Clinical Laboratory Standards agar dilution method was used to compare the in vitro activity of WY-49605 (also called SUN/SY 5555 and ALP-201), a new broad-spectrum oral penem, to those of amoxicillin, amoxicillin-clavulanate, imipenem, ciprofloxacin, cefaclor, cefpodoxime
S Furukawa et al.
The Japanese journal of antibiotics, 48(2), 210-219 (1995-02-01)
1. SY5555 dry syrup (powder which is dissolved before use) was administered to 25 patients with bacterial infections (6 cases of bronchitis, 2 cases of bronchopneumonia, 1 case of pertussis, 3 cases of scarlet fever, 5 cases of tonsillitis, 3
Treatment of acute uncomplicated cystitis with faropenem for 3 days versus 7 days: multicentre, randomized, open-label, controlled trial.
Hamasuna R, et al.
The Journal of Antimicrobial Chemotherapy, 69(6), 1675-1680 (2014)
J M Woodcock et al.
The Journal of antimicrobial chemotherapy, 39(1), 35-43 (1997-01-01)
The in-vitro activity of faropenem, a novel oral penem, was studied in comparison with other beta-lactam antimicrobials against 711 recent clinical isolates including Gram-negative, Gram-positive and anaerobic bacteria. MIC data showed that faropenem was active against most members of the

Articles

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service