Skip to Content
Merck
All Photos(3)

Key Documents

F6803

Sigma-Aldrich

D-Fructose 1,6-bisphosphate trisodium salt hydrate

≥98% (TLC)

Synonym(s):

D(+)Fructofuranose 1,6-diphosphate trisodium salt hydrate, Harden-Young ester, Hexose diphosphate trisodium salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11Na3O12P2 · xH2O
CAS Number:
Molecular Weight:
406.06 (anhydrous basis)
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

microbial

Quality Level

Assay

≥98% (TLC)

form

powder

technique(s)

thin layer chromatography (TLC): suitable

color

white to off-white

solubility

water: 50 mg/mL, clear, colorless to faintly yellow

cation traces

Na: 14.6-18.8% (dry basis)

storage temp.

−20°C

SMILES string

O.[Na+].[Na+].[Na+].O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H](O)C(=O)COP(O)([O-])=O

InChI

1S/C6H14O12P2.3Na.H2O/c7-3(1-17-19(11,12)13)5(9)6(10)4(8)2-18-20(14,15)16;;;;/h3,5-7,9-10H,1-2H2,(H2,11,12,13)(H2,14,15,16);;;;1H2/q;3*+1;/p-3/t3-,5-,6-;;;;/m1..../s1

InChI key

ISLNIFDAODOXHN-GNWSQLALSA-K

Looking for similar products? Visit Product Comparison Guide

Application

D-Fructose-1,6-bisphosphate (FBP), a common metabolic sugar, is the precursor of glyceraldehyde 3-phosphate and dihydroxyacetone phosphate in the glycolytic pathway. It may be used as an allosteric activator of enzymes such as pyruvate kinase and NAD+-dependent L-(+)-lactate dehydrogenase, as an inhibitor of acetate kinase and as a substrate to identify and characterize enzymes such as fructose-1,6-bisphosphate aldolase(s) and fructose-1,6-bisphosphatase(s). FBP is studied as a neuroprotective agent in brain injury.

Biochem/physiol Actions

Fructose-1,6-biphosphate (F1,6P) is a glycolytic intermediate produced by the transfer of a phosphate from ATP to fructose-6-phosphate by the enzyme phosphofructokinase. Fructose-1,6-biphosphate, along with fructose-2,6-biphosphate, modulates the activity of phosphofructokinase-1 (PFK-1), the rate-limiting step in glycolysis. Fructose-1,6-biphosphate is also an allosteric activator of the M2 isoform of Pyruvate Kinase (PK-M2), the predominant form of pyruvate kinase in cancer cells.
Fructose-1,6-biphosphate (F1,6P) is a glycolytic intermediate produced by the transfer of a phosphate from ATP to fructose-6-phosphate by the enzyme phosphofructokinase. Fructose-1,6-biphosphate, along with fructose-2,6-biphosphate, modulates the activity of phosphofructokinase-1 (PFK-1), the rate-limiting step in glycolysis. During glycolysis, aldolase splits Fructose-1,6-biphosphate into dihydroxacetone phosphate (DHAP) and glyceraldehyde phosphate. Fructose-1,6-biphosphate is also an allosteric activator of the M2 isoform of Pyruvate Kinase (PK-M2), the predominant form of pyruvate kinase in cancer cells.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Tao Yu et al.
Cell, 174(6), 1549-1558 (2018-08-14)
Engineering microorganisms for production of fuels and chemicals often requires major re-programming of metabolism to ensure high flux toward the product of interest. This is challenging, as millions of years of evolution have resulted in establishment of tight regulation of
Yasmean Kalam et al.
Clinical toxicology (Philadelphia, Pa.), 50(7), 546-554 (2012-08-09)
Fructose-1,6-diphosphate (FDP) is a metabolite in the glycolytic pathway created from glucose. Exogenously administered FDP increases the yield of ATP from anaerobic glycolysis. FDP reduces ischaemic tissue area in experimentally-induced cerebral and myocardial infarction and improves haemodynamics post-cardiac bypass. We
Roberto Christ Vianna Santos et al.
Inflammation, 35(3), 1198-1203 (2012-02-14)
It has been previously showed that fructose-1,6-bisphosphate (FBP) has anti-inflammatory and immunomodulatory effects on several experimental inflammation models. However, the effects and mechanism of FBP on Zymosan-induced acute lung injury (ALI) in mice had not been tested. In this study
Guilherme Vargas Bochi et al.
Inflammation, 35(6), 1786-1792 (2012-07-11)
The accumulation of advanced oxidation protein products (AOPP) has been linked to several pathological conditions. Previous studies have identified AOPP as a novel biomarker of oxidative damage to proteins and a novel class of mediator of inflammation. The aim of
Malkhey Verma et al.
Biochimica et biophysica acta, 1827(1), 19-29 (2012-10-04)
We develop a strategic 'domino' approach that starts with one key feature of cell function and the main process providing for it, and then adds additional processes and components only as necessary to explain provoked experimental observations. The approach is

Articles

Review the 10 steps of glycolysis in the Embden-Meyerhof-Parnas glycolytic pathway. Easily compare reaction stages and buy the enzymes for your life science research.

Review the 10 steps of glycolysis in the Embden-Meyerhof-Parnas glycolytic pathway. Easily compare reaction stages and buy the enzymes for your life science research.

Review the 10 steps of glycolysis in the Embden-Meyerhof-Parnas glycolytic pathway. Easily compare reaction stages and buy the enzymes for your life science research.

Review the 10 steps of glycolysis in the Embden-Meyerhof-Parnas glycolytic pathway. Easily compare reaction stages and buy the enzymes for your life science research.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service