Skip to Content
Merck
All Photos(2)

Key Documents

F5021

Sigma-Aldrich

Flunisolide

≥97%

Synonym(s):

6-Fluoro-11,16α,17,21-tetrahydroxypregna-1,4-diene-3,20-dione 16,17-acetonide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H31FO6
CAS Number:
Molecular Weight:
434.50
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Assay

≥97%

originator

Roche

SMILES string

CC1(C)O[C@@H]2CC3C4C[C@H](F)C5=CC(=O)C=CC5(C)C4C(O)CC3(C)[C@@]2(O1)C(=O)CO

InChI

1S/C24H31FO6/c1-21(2)30-19-9-14-13-8-16(25)15-7-12(27)5-6-22(15,3)20(13)17(28)10-23(14,4)24(19,31-21)18(29)11-26/h5-7,13-14,16-17,19-20,26,28H,8-11H2,1-4H3/t13?,14?,16-,17?,19+,20?,22?,23?,24+/m0/s1

InChI key

XSFJVAJPIHIPKU-CCRGDLLISA-N

Features and Benefits

This compound was developed by Roche. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 1 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carol R Reinero et al.
Journal of veterinary internal medicine, 20(1), 57-64 (2006-02-25)
Feline bronchial disease is commonly treated with oral glucocorticoids (OGC), which might be contraindicated in cats with certain infectious, endocrine, renal, or cardiac diseases. Inhalant GC (IGC) maximize local efficacy and minimize systemic bioavailability. We evaluated systemic endocrine and immune
T Schafer et al.
Rhinology, 49(3), 272-280 (2011-08-23)
Intranasal corticosteroids (INS) are the first line treatment for allergic rhinitis (AR). To guide clinical decision-making, we created a therapeutic index (TIX) for INS reflecting efficacy and safety. A Medline search (1966 to June 2009) was carried out to identify
F Decimo et al.
International journal of immunopathology and pharmacology, 22(2), 363-370 (2009-06-10)
The role of inhaled corticosteroids in asthma exacerbation is debated. We compared high doses of nebulized budesonide versus high doses of nebulized flunisolide, in association with a short-acting beta-2-agonist, in the treatment of moderate asthma exacerbation in preschool children. In
Yi Jin
Chemico-biological interactions, 191(1-3), 234-238 (2011-02-01)
Inhaled corticosteroids (ICS) are a mainstay anti-inflammatory therapy for the management of asthma. ICS are synthetic glucocorticoids that are structurally similar to the natural active human glucocorticoid cortisol. Steroid transforming enzymes of the aldo-keto reductase (AKR) family, namely AKR1D1 (5β-steroid
Jonathan Corren et al.
Clinical therapeutics, 25(3), 776-798 (2003-07-11)
Inhaled corticosteroids are currently recommended as first-line therapy for the long-term control and management of persistent asthma. Flunisolide hydrofluoroalkane (HFA) is a new formulation of the corticosteroid flunisolide that is delivered by a metered-dose inhaler containing an HFA propellant. HFA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service