Skip to Content
Merck
All Photos(2)

Key Documents

F5007

Sigma-Aldrich

Fibronectin Type III Connecting Segment Fragment 1-25

≥90% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C123H195N31O39
CAS Number:
Molecular Weight:
2732.05
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.32

biological source

synthetic (organic)

Quality Level

Assay

≥90% (HPLC)

form

powder

UniProt accession no.

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](NC(=O)[C@H](CCC(O)=O)NC(=O)[C@@H]1CCCN1C(=O)CNC(=O)[C@H](Cc2c[nH]cn2)NC(=O)[C@H](CC(C)C)NC(=O)[C@H](CC(N)=O)NC(=O)[C@@H]3CCCN3C(=O)[C@H](Cc4c[nH]cn4)NC(=O)[C@@H]5CCCN5C(=O)[C@H](CC(C)C)NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@H](CC(C)C)NC(=O)[C@H](CCC(N)=O)NC(=O)[C@@H]6CCCN6C(=O)[C@H](CC(C)C)NC(=O)[C@H](CCC(O)=O)NC(=O)[C@@H](N)CC(O)=O)C(C)C)[C@@H](C)O)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CC(O)=O)C(=O)N[C@@H](C(C)C)C(=O)N7CCC[C@H]7C(=O)N[C@@H](CO)C(=O)N[C@@H]([C@@H](C)O)C(O)=O

InChI

1S/C123H195N31O39/c1-19-64(16)97(117(186)140-74(42-58(4)5)106(175)138-78(50-94(167)168)109(178)146-96(63(14)15)122(191)154-40-24-29-87(154)115(184)144-82(54-155)110(179)149-99(66(18)157)123(192)193)147-104(173)72(32-35-92(163)164)134-111(180)83-25-20-36-150(83)90(160)53-129-101(170)76(46-67-51-127-55-130-67)137-105(174)73(41-57(2)3)136-107(176)77(49-89(126)159)139-113(182)85-27-22-39-153(85)121(190)81(47-68-52-128-56-131-68)142-114(183)86-28-23-38-152(86)120(189)80(45-61(10)11)143-118(187)98(65(17)156)148-116(185)95(62(12)13)145-108(177)75(43-59(6)7)135-102(171)70(30-33-88(125)158)133-112(181)84-26-21-37-151(84)119(188)79(44-60(8)9)141-103(172)71(31-34-91(161)162)132-100(169)69(124)48-93(165)166/h51-52,55-66,69-87,95-99,155-157H,19-50,53-54,124H2,1-18H3,(H2,125,158)(H2,126,159)(H,127,130)(H,128,131)(H,129,170)(H,132,169)(H,133,181)(H,134,180)(H,135,171)(H,136,176)(H,137,174)(H,138,175)(H,139,182)(H,140,186)(H,141,172)(H,142,183)(H,143,187)(H,144,184)(H,145,177)(H,146,178)(H,147,173)(H,148,185)(H,149,179)(H,161,162)(H,163,164)(H,165,166)(H,167,168)(H,192,193)/t64-,65+,66+,69-,70-,71-,72-,73-,74-,75-,76-,77-,78-,79-,80-,81-,82-,83-,84-,85-,86-,87-,95-,96-,97-,98-,99-/m0/s1

InChI key

NXAKDBCNODQXBZ-YSZCVPRFSA-N

Gene Information

human ... FN1(2335)

Amino Acid Sequence

Asp-Glu-Leu-Pro-Gln-Leu-Val-Thr-Leu-Pro-His-Pro-Asn-Leu-His-Gly-Pro-Glu-Ile-Leu-Asp-Val-Pro-Ser-Thr

General description

Fibronectin is composed of two cell adhesion domains namely, the central domain for cell-binding made of adhesion-recognizing tetrapeptide, and the alternatively spliced type III connecting segment (IIICS). It contains a tetrapeptide region, from amino acid 90-109, called CS5, which is essential for adhesion mediated by IIICS. Both CS5 and CS1 mediate melanoma cell spreading, when stimulated by immunoglobulin (Ig)G, with CS1 exhibiting higher activity.

Biochem/physiol Actions

Fibronectin Type III Connecting Segment (IIICS) is responsible for melanoma cell adhesion. It is the predominant binding site of integrin α4β1, which is mediated through the aspartic acid residue present at position 21. Studies in chicken show that alternative splicing in this region during development of the peripheral nervous system, is essential for the control cell-cell adhesion.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Vanderslice et al.
Journal of immunology (Baltimore, Md. : 1950), 158(4), 1710-1718 (1997-02-15)
The alpha 4 integrins mediate leukocyte adhesion to specific counter-receptors, including vascular cell adhesion molecule-1 (VCAM-1), the fibronectin splice variant containing connecting segment 1 (CS1), and mucosal addressin cell adhesion molecule-1. A series of cyclized peptides based on the LDV
Kosuke Azuma et al.
Arteriosclerosis, thrombosis, and vascular biology, 26(10), 2275-2280 (2006-08-05)
The aim of this study was to elucidate the effect of repetitive fluctuations in blood glucose concentrations on monocyte adhesion to the aortic endothelium. Nonobese type 2 diabetes, Goto-Kakizaki (GK) rats were fed twice daily to induce repetitive postprandial glucose
I N Jongewaard et al.
Cell adhesion and communication, 3(6), 487-495 (1996-04-01)
The type III connecting segment (IIICS) within fibronectin is the major binding site for the integrin alpha 4 beta 1. Most integrin ligands have an essential acidic residue within their integrin binding site, in IIICS this residue is hypothesized to
C F Craddock et al.
British journal of haematology, 97(1), 15-21 (1997-04-01)
In vitro the integrin VLA4 mediates the adhesion of haemopoietic progenitors to bone marrow stroma through an interaction with its ligands VCAM-1 and the CS1 moiety of fibronectin. The VLA4/VCAM-1 pathway has been implicated in haemopoietic trafficking in vivo since
Pachiyappan Kamarajan et al.
BMC cancer, 10, 330-330 (2010-06-29)
The alternatively spliced V region or type III connecting segment III (IIICS) of fibronectin is important in early development, wound healing, and tumorigenesis, however, its role in oral cancer has not been fully investigated. Thus, we investigated the role of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service