Skip to Content
Merck
All Photos(1)

Key Documents

F3885

Sigma-Aldrich

Fetal Bovine Serum

γ-irradiated, USA origin, sterile-filtered, suitable for cell culture, suitable for hybridoma

Synonym(s):

GI FBS, GI FCS, GI Fetal Bovine Sera, GI Fetal Bovine Serum, Gamma Irradiated Fetal Calf Sera, FBS, FCS, sera, serum

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.71
biological source:
bovine fetus
origin:
USA origin
sterility:
γ-irradiated
sterile-filtered
shipped in:
dry ice

biological source

bovine fetus

sterility

γ-irradiated
sterile-filtered

product line

FBS Classic

origin

USA origin

technique(s)

cell culture | hybridoma: suitable
cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Preparation Note

Prepared by exposure to a 60Co source.

Analysis Note

  • Endotoxin and hemoglobin tested
  • Tested for the presence of bacteria, virus, and mycoplasma
  • Triple filtered with 0.1 micron membrane under aseptic conditions

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shabir Ahmad Lone et al.
BMC complementary medicine and therapies, 20(1), 131-131 (2020-05-01)
The global prevalence of fungal diseases is increasing rapidly, which affects more than a billion people every year with significant mortality rate. On the other hand, the development of new drugs to treat these fungal infections is slow, while the

Articles

Find answers to questions (FAQs) about fetal bovine serum (FBS) that is gamma-irradiated by the SER-TAIN™ process. Request a sample of FBS at Sigma-Aldrich.com.

Find answers to questions (FAQs) about fetal bovine serum (FBS) that is gamma-irradiated by the SER-TAIN™ process. Request a sample of FBS at Sigma-Aldrich.com.

Find answers to questions (FAQs) about fetal bovine serum (FBS) that is gamma-irradiated by the SER-TAIN™ process. Request a sample of FBS at Sigma-Aldrich.com.

Find answers to questions (FAQs) about fetal bovine serum (FBS) that is gamma-irradiated by the SER-TAIN™ process. Request a sample of FBS at Sigma-Aldrich.com.

Protocols

Protect your investment with best practices for storing and thawing FBS and other sera. Sample FBS for validation in your cell culture protocol.

Protect your investment with best practices for storing and thawing FBS and other sera. Sample FBS for validation in your cell culture protocol.

Related Content

Explore our portfolio of fetal bovine serum (FBS) and determine the right FBS for your lab needs and budget. Request a serum sample.

Explore our portfolio of fetal bovine serum (FBS) and determine the right FBS for your lab needs and budget. Request a serum sample.

Explore our portfolio of fetal bovine serum (FBS) and determine the right FBS for your lab needs and budget. Request a serum sample.

Explore our portfolio of fetal bovine serum (FBS) and determine the right FBS for your lab needs and budget. Request a serum sample.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service