Skip to Content
Merck
All Photos(1)

Key Documents

F3256

Sigma-Aldrich

Fetuin-Agarose

saline suspension

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.56

biological source

bovine blood (Fetal)

Quality Level

form

saline suspension

extent of labeling

≥200 μg per mL

technique(s)

affinity chromatography: suitable

matrix

cross-linked 4% beaded agarose

matrix activation

cyanogen bromide

matrix attachment

amino

matrix spacer

1 atom

suitability

suitable for chromatography

storage temp.

2-8°C

Application

Fetuin-agarose is an agarose conjugate in saline suspension used in affinity chromatography, protein chromatography and specialty resins. Fetuin-agarose columns have been used in studies that inform the detection and quantification of tumor markers.

Physical form

Suspension in 0.5M NaCl containing preservative

Preparation Note

Prepared with Fetuin, F2379

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M J Friedman
Proceedings of the National Academy of Sciences of the United States of America, 80(17), 5421-5424 (1983-09-01)
During malaria and other infections, the plasma concentration of alpha 1-acid glycoprotein (AGP) increases 3- to 4-fold, but the function of this glycoprotein has been unknown. This study demonstrates, by in vitro culture of the malaria parasite Plasmodium falciparum, that
Nohora Vega et al.
Phytochemistry, 67(4), 347-355 (2006-01-18)
A lectin was isolated and characterised from Salvia bogotensis seeds. Removal of the abundant pigments and polysaccharides, which are present in seeds, was an essential step in its purification. Several procedures were assayed and the best suited, including Pectinex treatment
G D Armstrong et al.
Infection and immunity, 55(5), 1294-1299 (1987-05-01)
We developed a method to produce radioiodinated pertussis toxin (PT) which was active in the goose erythrocyte agglutination and CHO cell assay systems. The procedure used fetuin coupled to agarose to prevent inactivation of the toxin during the iodination reaction.
Eduardo Armienta-Aldana et al.
Pakistan journal of biological sciences : PJBS, 12(5), 459-462 (2009-07-08)
We extracted and partial characterized lectin from runner beans (Phaseolus coccineus L.). This lectin shows a great affinity to fetuin-agarose column like others lectins and the electrophoretic gels point one band of approximately 45 kDa. In addition to the previous
Specificity of the <I>Vateairea macrocarpa</I> lectin towards glycans exhibiting exposed Gal/GalNAc residues
Ramos, M.V., et al.
Protein and peptide letters, 6(3), 163-171 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service