Skip to Content
Merck
All Photos(3)

Key Documents

F114

Sigma-Aldrich

cis-(Z)-Flupenthixol dihydrochloride

≥98% (HPLC), solid

Synonym(s):

(Z)-4-[3-[2-(Trifluoromethyl)-9H-thioxanthen-9-ylidene]propyl]-1-piperazineethanol dihydrochloride, Flupentixol dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H25F3N2OS · 2HCl
CAS Number:
Molecular Weight:
507.44
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

solid

color

white to off-white

solubility

H2O: soluble

originator

Lundbeck

SMILES string

Cl[H].Cl[H].OCCN1CCN(CC\C=C2\c3ccccc3Sc4ccc(cc24)C(F)(F)F)CC1

InChI

1S/C23H25F3N2OS.2ClH/c24-23(25,26)17-7-8-22-20(16-17)18(19-4-1-2-6-21(19)30-22)5-3-9-27-10-12-28(13-11-27)14-15-29;;/h1-2,4-8,16,29H,3,9-15H2;2*1H/b18-5-;;

InChI key

IOVDQEIIMOZNNA-MHKBYHAFSA-N

General description

cis-(Z)-Flupenthixol dihydrochloride (FLU) is a thioxanthene drug.[1] DRD1 (dopamine receptor D1) codes for D1Rs and is located on human chromosome 5q35.2.[2] Dopamine D1 is expressed on lung alveolar type I cells.[3]

Application

cis-(Z)-Flupenthixol dihydrochloride has been used in drug infusions.[4] It has been used to determine the causal involvement of extracellular LS-DA and LS-NE release patterns on social play behavior.[5]

Biochem/physiol Actions

Antipsychotic; neuroleptic agent; dopamine receptor antagonist. cis-(Z)-Flupenthixol dihydrochloride (FLU) is used to treat schizophrenia and anxiolytic and depressive disorders[1] Dopamine D1 plays a major role in lung fluid homeostasis.[3] It controls the airway smooth muscle tone by producing adenylyl cyclase/cAMP, that would favor airway relaxation in asthmatics.[3] Dopamine in the lung, serves as a neurotransmitter and noradrenaline precursor.[3]

Features and Benefits

This compound was developed by Lundbeck. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Legal Information

Sold with the permission of H. Lundbeck A/S.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Involvement of dopamine, but not norepinephrine, in the sex-specific regulation of juvenile socially rewarding behavior by vasopressin
Bredewold R, et al.
Neuropsychopharmacology (2018)
Transcriptomic context of DRD1 is associated with prefrontal activity and behavior during working memory
Fazio L, et al.
Proceedings of the National Academy of Sciences of the USA, 115(21), 5582-5587 (2018)
The dopamine D 1 receptor is expressed and induces CREB phosphorylation and MUC5AC expression in human airway epithelium
Matsuyama N, et al.
Respiratory Research, 19(1), 53-53 (2018)
Positive reinforcement mediated by midbrain dopamine neurons requires D1 and D2 receptor activation in the nucleus accumbens
Steinberg E E, et al.
PLoS ONE, 9(4), e94771-e94771 (2014)
Prolonged neuroleptic effect of alpha-flupenthixol decanoate in oil.
M Nymark et al.
Acta pharmacologica et toxicologica, 33(5), 363-376 (1973-01-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service