Skip to Content
Merck
All Photos(1)

Key Documents

EPS002

Sigma-Aldrich

MS-275

A HDAC1 and HDAC3 inhibitor

Synonym(s):

MS-275 (Entinostat, SNDX-275), 3-pyridinylmethyl [[4-[[(2-aminophenyl)amino]carbonyl]phenyl]methyl]carbamate, N-(2-Aminophenyl)-4-[N-(pyridine-3ylmethoxycarbonyl)aminomethyl]benzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H20N4O3
CAS Number:
Molecular Weight:
376.41
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.41

Assay

≥99%

solubility

DMSO: 38 mg/mL

storage temp.

−20°C

SMILES string

Nc1ccccc1NC(=O)c2ccc(CNC(=O)OCc3cccnc3)cc2

InChI

1S/C21H20N4O3/c22-18-5-1-2-6-19(18)25-20(26)17-9-7-15(8-10-17)13-24-21(27)28-14-16-4-3-11-23-12-16/h1-12H,13-14,22H2,(H,24,27)(H,25,26)

InChI key

INVTYAOGFAGBOE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Preferentially inhibits HDAC1 (IC50=300 nM) over HDAC3 (IC50=8 μM). Has no inhibitory activity towards HDAC8 (IC50>100 μM).

Application

MS-275 has been used as a histone deacetylase-1 inhibitor.

Biochem/physiol Actions

HDAC inhibitor; antiproliferative.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Repr. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heather MacTavish et al.
PloS one, 5(12), e14462-e14462 (2011-02-02)
Histone deacetylase inhibitors (HDI) dampen cellular innate immune response by decreasing interferon production and have been shown to increase the growth of vesicular stomatitis virus and HSV. As attenuated tumour-selective oncolytic vaccinia viruses (VV) are already undergoing clinical evaluation, the
Marta Bombardo et al.
Scientific reports, 8(1), 9391-9391 (2018-06-22)
Adult pancreatic acinar cells have the ability to re-enter the cell cycle and proliferate upon injury or tissue loss. Despite this mitotic ability, the extent of acinar proliferation is often limited and unable to completely regenerate the injured tissue or
HoxC5 and miR-615-3p target newly evolved genomic regions to repress hTERT and inhibit tumorigenesis
TingDong Y et al.
Nature Communications, 100 (2018)
Guido Dentesano et al.
Journal of neuroinflammation, 9, 165-165 (2012-07-11)
In physiological conditions, it is postulated that neurons control microglial reactivity through a series of inhibitory mechanisms, involving either cell contact-dependent, soluble-factor-dependent or neurotransmitter-associated pathways. In the current study, we focus on CD200R1, a microglial receptor involved in one of
Ilaria Lepore et al.
PloS one, 8(12), e83018-e83018 (2013-12-19)
Over the past years BARD1 (BRCA1-associated RING domain 1) has been considered as both a BRCA1 (BReast Cancer susceptibility gene 1, early onset) interactor and tumor suppressor gene mutated in breast and ovarian cancers. Despite its role as a stable

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service