Skip to Content
Merck
All Photos(1)

Key Documents

EP5405000107

Eppendorf® Centrifuge 5425

refrigerated: no, AC/DC input 120 V, 50 - 60 Hz, US 3-pin plug

Synonym(s):

Eppendorf® Centrifuge 5425

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161701

feature

CE compliant
CSA approved
refrigerated: no

packaging

pack of 1 ea

manufacturer/tradename

Eppendorf® 5405000107

technique(s)

centrifugation: suitable

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Extended Rim rotor for increased safety. Quicklock for fast and easy operation. Whisper quiet.
Centrifuge 5425 is the new laboratory standard with its 24-place capacity and speed up to 21, 330 x G. Focusing on standard applications, it is an essential piece of equipment for modern labs ensuring optimal separation. The centrifuge 5425 serves as a reliable Partner especially for high-throughput users Expecting a top level of ergonomic comfort and provides great flexibility with its 6 rotor options.

Application

Centrifugation

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service