EP4967000014
Eppendorf® Varispenser® 2x Bottle Top Dispenser
0.2-2 mL, with recirculation valve, with telescopic intake tube (length 125 – 240 mm)
About This Item
Recommended Products
feature
autoclavable
incremental 0.05 mL
comes with adapters GL 25, GL 28/S 28, GL 32, GL 38, S 40
with recirculation valve
with telescopic intake tube (length 125 – 240 mm)
manufacturer/tradename
Eppendorf® 4967000014
measuring range
0.2-2 mL
volume
0.2-2 mL
fitting
(GL 28/S 28)
GL 25
GL 32
GL 38
S 40
Looking for similar products? Visit Product Comparison Guide
General description
Product Information
Varispenser 2/2x are the ideal choice for dispensing aliquots of liquid from supply and reagent bottles. Advanced sealing lip technology enables dispensing of nearly all kinds of acids, solutions or bases. Available in 6 sizes for dispensing volumes from 0.2 – 100 mL, these instruments are designed to consistently dispense selected volumes without reagent waste. All Varispenser 2/2x dispensers are fully autoclavable for maximum safety in your lab. The Varispenser 2x is additionally equipped with a recirculation valve for priming the dispensing system without loss of reagent. The 360 ° rotating valve block offers safe access in all positions.
For laboratory tasks with aliquots of aggressive solutions like lyes, acids, bases or solvents taken from large supply and reagent bottles, bottle top dispensers need to be universally applicable and of uncompromising quality. The new Varispenser 2 and Varispenser 2x bottle top dispensers offer high chemical resistance and robustness in the range of volumes up to 100 mL. Varispenser 2 x is additionally equipped with a recirculation valve for priming the dispensing system without loss of reagent. The trusted PFA sealing lip technology prevents crystal building from media residue by wiping off the inner cylinder wall during dispensing. This technology combined with the high chemical resistance allows the use with nearly all kinds of liquids. The extra air vent cap with Luer-cone for the assembly of a microfilter prevents possible contamination from the supply bottle. Varispenser 2 and Varispenser 2x are completely autoclavable. The 360 ° rotating valve block offers safe and easy access in all positions. Because of the GL 45 thread on all sizes, Varispenser 2 and Varipsenser 2x can be used on most common bottle threads in the laboratory. Depending on the chosen size, Varispenser 2 and Varispenser 2x come with 3 or 5 additional thread adapters
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service