Skip to Content
Merck
All Photos(1)

Key Documents

EHU184131

Sigma-Aldrich

MISSION® esiRNA

targeting human SOX2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GTTTGCAAAAGGGGGAAAGTAGTTTGCTGCCTCTTTAAGACTAGGACTGAGAGAAAGAAGAGGAGAGAGAAAGAAAGGGAGAGAAGTTTGAGCCCCAGGCTTAAGCCTTTCCAAAAAATAATAATAACAATCATCGGCGGCGGCAGGATCGGCCAGAGGAGGAGGGAAGCGCTTTTTTTGATCCTGATTCCAGTTTGCCTCTCTCTTTTTTTCCCCCAAATTATTCTTCGCCTGATTTTCCTCGCGGAGCCCTGCGCTCCCGACACCCCCGCCCGCCTCCCCTCCTCCTCTCCCCCCGCCCGCGGGCCCCCCAAAGTCCCGGCCGGGCCGAGGGTCGGCGGCCGCCGGCGGGCCGGGCCCGCGCACAGCGCCCGCATGTACAACATGATGGAGACGGAGCTGAA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

human ... SOX2(6657) , Sox2()

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liang Tang et al.
Pathology oncology research : POR, 24(4), 907-913 (2018-04-06)
Osteosarcoma (OS) was a prevalent malignant bone tumor which threatens people's health worldwide. Wnt/β catenin signaling pathway had been proved significant in various cancers, indicating its possible function in OS as well. Sox2, a crucial member among SOX family could
Jiaxuan Chen et al.
Molecular carcinogenesis, 56(10), 2267-2278 (2017-05-26)
Fas signaling promotes colorectal cancer (CRC) metastasis by inducing epithelial-mesenchymal transition (EMT). The acquisition of EMT properties in turn induces stemness but the mechanism by which Fas signaling contributes to it still remains unclear. Hence, the aim of this study
Tatsuya Ishiguro et al.
Cancer research, 76(1), 150-160 (2015-12-17)
The establishment of cancer stem-like cell (CSC) culture systems may be instrumental in devising strategies to fight refractory cancers. Inhibition of the Rho kinase ROCK has been shown to favorably affect CSC spheroid cultures. In this study, we show how
Nicolas Chassaing et al.
Genome research, 26(4), 474-485 (2016-02-20)
Ocular developmental anomalies (ODA) such as anophthalmia/microphthalmia (AM) or anterior segment dysgenesis (ASD) have an estimated combined prevalence of 3.7 in 10,000 births. Mutations in SOX2 are the most frequent contributors to severe ODA, yet account for a minority of
Keshav Gopal et al.
Oncotarget, 7(3), 3111-3127 (2015-12-20)
We have previously identified a novel intra-tumoral dichotomy in breast cancer based on the differential responsiveness to a Sox2 reporter (SRR2), with cells responsive to SRR2 (RR) being more stem-like than unresponsive cells (RU). Here, we report that RR cells

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service