Skip to Content
Merck
All Photos(1)

Key Documents

EHU113921

Sigma-Aldrich

MISSION® esiRNA

targeting human ATG4B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GGCAGCCAGACAGCTACTTCAGCGTCCTCAACGCATTCATCGACAGGAAGGACAGTTACTACTCCATTCACCAGATAGCGCAAATGGGAGTTGGCGAAGGCAAGTCCATAGGCCAGTGGTACGGGCCCAACACTGTCGCCCAGGTCCTGAAGAAGCTTGCTGTCTTCGATACGTGGAGCTCCTTGGCGGTCCACATTGCAATGGACAACACTGTTGTGATGGAGGAAATCAGAAGGTTGTGCAGGACCAGCGTTCCCTGTGCAGGCGCCACTGCGTTTCCTGCAGATTCCGACCGGCACTGCAACGGATTCCCTGCCGGAGCTGAGGTCACCAACAGGCCGTCGCCATGGAGACCCCTGGTACTTCTCATTCCCCTGCGCCTGGGGCTCACGGACATCAACGAGGCCTACGT

Ensembl | human accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yao-Xin Lin et al.
Biomaterials, 141, 199-209 (2017-07-10)
Autophagic therapy is regarded as a promising strategy for disease treatment. Appropriate autophagy regulations in vivo play a crucial role in translating this new concept from benchside to bedside. So far, emerging technologies are required to spatially and quantitatively monitor autophagic
Pei-Feng Liu et al.
Cancers, 11(12) (2019-11-28)
Oral squamous cell carcinoma (OSCC) is one of the major leading causes of cancer death worldwide due to the limited availability of biomarkers and therapeutic targets. Autophagy related protease 4B (ATG4B) is an essential protease for the autophagy machinery, and
Tianzhi Huang et al.
Cancer cell, 32(6), 840-855 (2017-12-13)
ATG4B stimulates autophagy by promoting autophagosome formation through reversible modification of ATG8. We identify ATG4B as a substrate of mammalian sterile20-like kinase (STK) 26/MST4. MST4 phosphorylates ATG4B at serine residue 383, which stimulates ATG4B activity and increases autophagic flux. Inhibition
Pei-Feng Liu et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 44(2), 728-740 (2017-11-24)
ATG4B is a cysteine protease required for autophagy, which is a cellular catabolic pathway involved in energy balance. ATG4B expression is elevated during tumor growth in certain types of cancer, suggesting that ATG4B is an attractive target for cancer therapy.
Yao-Xin Lin et al.
ACS nano, 11(2), 1826-1839 (2017-01-24)
Autophagy plays a crucial role in the metabolic process. So far, conventional methods are incapable of rapid, precise, and real-time monitoring of autophagy in living objects. Herein, we describe an in situ intracellular self-assembly strategy for quantitative and temporal determination

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service