Skip to Content
Merck
All Photos(1)

Key Documents

EHU068551

Sigma-Aldrich

MISSION® esiRNA

targeting human YTHDF2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

ACTTGAGTCCACAGGCAAGGCCCAATAATGCATATACTGCCATGTCAGATTCCTACTTACCCAGTTACTACAGTCCCTCCATTGGCTTCTCCTATTCTTTGGGTGAAGCTGCTTGGTCTACGGGGGGTGACACAGCCATGCCCTACTTAACTTCTTATGGACAGCTGAGCAACGGAGAGCCCCACTTCCTACCAGATGCAATGTTTGGGCAACCAGGAGCCCTAGGTAGCACTCCATTTCTTGGTCAGCATGGTTTTAATTTCTTTCCCAGTGGGATTGACTTCTCAGCATGGGGAAATAACAGTTCTCAGGGACAGTCTACTCAGAGCTCTGGATATAGTAGCAATTATGCTTATGCACCTAGCTCCTTAGGTGGAGCCATGATTGATGGACAGTCAGCTTTTGCCAATGA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chuanzhao Zhang et al.
Oncogene, 39(23), 4507-4518 (2020-05-06)
N6-methyladenosine (m6A) RNA methylation contributes to the cancer stem cell (CSC) phenotype through regulating gene expression. YTHDF2, an m6A reader, was shown to be associated with hepatocellular carcinoma (HCC) patient prognosis. However, the effect of YTHDF2 on liver CSC and
Ruifan Wu et al.
Biochimica et biophysica acta. Gene regulatory mechanisms, 1862(8), 796-806 (2019-07-12)
N6-methyladenosine (m6A), the most abundant internal mRNA modification in eukaryotes, plays a vital role in regulating adipogenesis. However, its underlying mechanism remains largely unknown. Here, we reveal that deletion of m6A demethylase FTO in porcine and mouse preadipocytes inhibits adipogenesis
Sara Zaccara et al.
Cell, 181(7), 1582-1595 (2020-06-04)
N6-methyladenosine (m6A) is the most abundant mRNA nucleotide modification and regulates critical aspects of cellular physiology and differentiation. m6A is thought to mediate its effects through a complex network of interactions between different m6A sites and three functionally distinct cytoplasmic
Ye Fu et al.
Nature chemical biology, 16(9), 955-963 (2020-05-27)
Diverse RNAs and RNA-binding proteins form phase-separated, membraneless granules in cells under stress conditions. However, the role of the prevalent mRNA methylation, m6A, and its binding proteins in stress granule (SG) assembly remain unclear. Here, we show that m6A-modified mRNAs
Gaocai Li et al.
Cell death & disease, 11(2), 103-103 (2020-02-08)
N6 methyladenosine (m6A) is one of the most prevalent epitranscriptomic modifications of mRNAs, and plays a critical role in various bioprocesses. Bone-derived mesenchymal stem cells (BMSCs) can attenuate apoptosis of nucleus pulposus cells (NPCs) under compression; however, the underlying mechanisms

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service