Skip to Content
Merck
All Photos(1)

Key Documents

EHU043821

Sigma-Aldrich

MISSION® esiRNA

targeting human RB1CC1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

ATGGCCAAGATTCCACTGTTGGAGTGCCTAACCAGACATAGTTACAGAGAATGTTTGGGAAGACTGGATTCTTTACCTGAACATGAAGACTCAGAAAAAGCTGAGATGAAAAGATCCACTGAACTGGTGCTCTCTCCTGATATGCCTAGAACAACTAACGAATCTTTGTTAACCTCATTTCCCAAGTCAGTGGAACATGTGTCCCCAGATACCGCAGATGCTGAAAGTGGCAAAGAAATTAGGGAATCTTGTCAAAGTACTGTTCATCAGCAAGATGAAACTACGATTGACACTAAAGATGGTGATCTGCCCTTTTTTAATGTCTCTTTGTTAGACTGGATAAATGTTCAAGATAGACCTAATGATGTGGAATCTTTGGTCAGGAAGTGCTTTGATTCTATGAGCAGGCTTGATCCAAGGATTATTCGACCATTTATAGCAGAATGCCGTCAAACTATTGCC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ingrid Kjos et al.
EMBO reports, 18(10), 1727-1739 (2017-08-25)
Autophagy (macroautophagy) is a highly conserved eukaryotic degradation pathway in which cytosolic components and organelles are sequestered by specialized autophagic membranes and degraded through the lysosomal system. The autophagic pathway maintains basal cellular homeostasis and helps cells adapt during stress;
Aykut Turan et al.
The Journal of cell biology, 218(2), 508-523 (2018-12-28)
Dendritic cells (DCs) are crucial for the induction of potent antiviral immune responses. In contrast to immature DCs (iDCs), mature DCs (mDCs) are not permissive for infection with herpes simplex virus type 1 (HSV-1). Here, we demonstrate that HSV-1 infection
Gaocai Li et al.
Cell death & disease, 11(2), 103-103 (2020-02-08)
N6 methyladenosine (m6A) is one of the most prevalent epitranscriptomic modifications of mRNAs, and plays a critical role in various bioprocesses. Bone-derived mesenchymal stem cells (BMSCs) can attenuate apoptosis of nucleus pulposus cells (NPCs) under compression; however, the underlying mechanisms
Eleonora Turco et al.
Molecular cell, 74(2), 330-346 (2019-03-12)
The autophagy cargo receptor p62 facilitates the condensation of misfolded, ubiquitin-positive proteins and their degradation by autophagy, but the molecular mechanism of p62 signaling to the core autophagy machinery is unclear. Here, we show that disordered residues 326-380 of p62
Takashi Nozawa et al.
Nature communications, 11(1), 770-770 (2020-02-09)
Invading microbial pathogens can be eliminated selectively by xenophagy. Ubiquitin-mediated autophagy receptors are phosphorylated by TANK-binding kinase 1 (TBK1) and recruited to ubiquitinated bacteria to facilitate autophagosome formation during xenophagy, but the molecular mechanism underlying TBK1 activation in response to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service