Skip to Content
Merck
All Photos(1)

Key Documents

E8156

Sigma-Aldrich

Anti-eRF1 antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Eukaryotic translation termination factor 1, Anti-RF1, Anti-Release factor 1, Anti-eTF1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 50 kDa

species reactivity

rat, human, mouse

packaging

antibody small pack of 25 μL

concentration

~1 mg/mL

technique(s)

immunoprecipitation (IP): 1-2 μg using HeLa cell lysates.
western blot: 0.5-1 μg/mL using 3T3 or Rat1 cell extracts.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ETF1(2107)
mouse ... Etf1(225363)
rat ... Etf1(307503)

General description

Eukaryotic translation termination factor 1 (eRF1) is a protein encoded by the ETF1 gene in humans. It belongs to eRF1 family and the amino-acid sequence is highly conserved.

Application

Anti-eRF1 antibody produced in rabbit has been used in western blotting and immunoprecipitation.

Biochem/physiol Actions

Eukaryotic translation termination factor 1 (eRF1) proteins are directly implicated in the termination of translation in eukaryotes. It plays an important role in the termination of protein synthesis, which is performed by three domains (N, M and C) of eRF1. Rearrangement in eRF1 leads to N-domain accommodation into the ribosomal A site.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cosuppression of eukaryotic release factor 1-1 in Arabidopsis affects cell elongation and radial cell division
Petsch K, et al.
Plant Physiology, 139(1), 115-126 (2005)
Identification and characterization of a novel ubiquitous nucleolar protein `NARR?encoded by a gene overlapping the rab34 oncogene
Zougman A, et al.
Nucleic Acids Research, 39(16), 7103-7113 (2011)
L Frolova et al.
Nature, 372(6507), 701-703 (1994-12-15)
The termination of protein synthesis in ribosomes is governed by termination (stop) codons in messenger RNAs and by polypeptide chain release factors (RFs). Although the primary structure of prokaryotic RFs and yeast mitochrondrial RF is established, that of the only
Toshiaki Shigeoka et al.
Nucleic acids research, 40(14), 6887-6897 (2012-05-05)
Upf1 is a highly conserved RNA helicase essential for nonsense-mediated mRNA decay (NMD), an mRNA quality-control mechanism that degrades aberrant mRNAs harboring premature termination codons (PTCs). For the activation of NMD, UPF1 interacts first with a translation-terminating ribosome and then
Tatsuaki Kurosaki et al.
Nature structural & molecular biology, 25(10), 940-950 (2018-10-03)
Nonsense-mediated messenger RNA decay (NMD) controls mRNA quality and degrades physiologic mRNAs to fine-tune gene expression in changing developmental or environmental milieus. NMD requires that its targets are removed from the translating pool of mRNAs. Since the decay steps of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service