Skip to Content
Merck
All Photos(1)

Key Documents

E3403

Millipore

EZview Red Protein G Affinity Gel

Synonym(s):

EZview Affinity Gel, Protein G Affinity Gel

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.56

form

liquid

analyte chemical class(es)

proteins (Immunoglobulins of various mammalian species)

technique(s)

affinity chromatography: suitable
immunoprecipitation (IP): suitable

capacity

≥8 mg/mL, gel binding capacity (rabbit IgG)

storage temp.

−20°C

General description

EZ view Red Protein G Affinity Gel is Protein G covalently bonded to 4% Agarose beads. The affinity gel is used to bind the FC portion of the IgG of an antibody to samples, such as cell lysates and tissue, for purification of proteins in preparation of immunoprecipitation assays. Red dye enhances visability for more efficient results.

Application

When performing small-scale affinity capture, such as immunoprecipitation, the affinity matrix is difficult to see in the microcentrifuge tubes. Accidental aspiration of the resin leads to quantitative variability in results. The EZview Red Affinity Gels greatly reduce the risk of pellet loss. EZview resins perform as well as conventional non-colored affinity gels, but allow the user to easily differentiate pellet from supernatant. This correlates to more accurate data because less protein is lost.

Features and Benefits

  • Increased visibility - Red color reduces risk of incidental aspiration
  • Improved recovery of target protein by reduced accidental loss
  • Higher reproducibility - More consistent yields

Physical form

1:1 (v/v) suspension in PBS containing 50% glycerol and 15 ppm Kathon
50% slurry in PBS with 50%Glycerol and 0.0015%

Legal Information

EZview is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Min Mo et al.
Proceedings of the National Academy of Sciences of the United States of America, 106(46), 19527-19532 (2009-11-06)
The anaphase-promoting complex (APC), or cyclosome, is a ubiquitin ligase with major roles in cell cycle regulation. It is required for mitotic exit, but must be deactivated for the G(1)/S phase transition to occur. APC consists of at least 12
Yuval Malka et al.
Nature communications, 8(1), 2029-2029 (2017-12-13)
The majority of mammalian genes contain one or more alternative polyadenylation sites. Choice of polyadenylation sites was suggested as one of the underlying mechanisms for generating longer/shorter transcript isoforms. Here, we demonstrate that mature mRNA transcripts can undergo additional cleavage and
Xi Lin et al.
The Journal of biological chemistry, 295(33), 11764-11775 (2020-06-27)
Macrophages play critical roles in homeostasis and inflammation. Macrophage polarization to either a pro-inflammatory or anti-inflammatory status is controlled by activating inflammatory signaling pathways. Ubiquitination is a posttranslational modification that regulates these inflammatory signaling pathways. However, the influence of protein
Juri Kazakevych et al.
Nucleic acids research, 45(10), 5770-5784 (2017-03-24)
Epigenetic mechanisms, including chromatin structure, chromatin dynamics and histone modifications play an important role for maintenance and differentiation of pluripotent embryonic stem cells. However, little is known about the molecular mechanisms of adult stem cell specification and differentiation. Here, we
Alvaro Quintanal-Villalonga et al.
Lung cancer (Amsterdam, Netherlands), 131, 112-121 (2019-04-28)
Lung adenocarcinoma accounts for approximately half of lung cancer cases. Twenty to 50% of tumors of this type harbor mutations affecting epidermal growth factor receptor (EGFR) expression or activity, which can be therapeutically targeted. EGFR inhibitors in this context exhibit

Related Content

Access high-quality reagents for antibody workflows, including adjuvants, blocking reagents, conjugation kits, and more.

Access high-quality reagents for antibody workflows, including adjuvants, blocking reagents, conjugation kits, and more.

Access high-quality reagents for antibody workflows, including adjuvants, blocking reagents, conjugation kits, and more.

Access high-quality reagents for antibody workflows, including adjuvants, blocking reagents, conjugation kits, and more.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service