Skip to Content
Merck
All Photos(3)

Documents

E3375

Sigma-Aldrich

trans-Androsterone

Synonym(s):

3β-Hydroxy-5α-androstan-17-one, 3β-Hydroxyetioallocholan-17-one, 5α-Androstan-3β-ol-17-one, Epi-androsterone, Isoandrosterone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H30O2
CAS Number:
Molecular Weight:
290.44
Beilstein:
1884007
EC Number:
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Quality Level

sterility

non-sterile

Assay

≥98% (HPLC)

form

powder

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

solubility

chloroform: 50 mg/mL, clear to slightly hazy, colorless to faintly yellow

shipped in

ambient

storage temp.

room temp

SMILES string

[H][C@@]12CC[C@@]3([H])[C@]4([H])CCC(=O)[C@@]4(C)CC[C@]3([H])[C@@]1(C)CC[C@H](O)C2

InChI

1S/C19H30O2/c1-18-9-7-13(20)11-12(18)3-4-14-15-5-6-17(21)19(15,2)10-8-16(14)18/h12-16,20H,3-11H2,1-2H3/t12-,13-,14-,15-,16-,18-,19-/m0/s1

InChI key

QGXBDMJGAMFCBF-LUJOEAJASA-N

Gene Information

human ... HSD17B3(3293)

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christopher C Waller et al.
Drug testing and analysis, 12(6), 752-762 (2020-01-18)
Hemapolin (2α,3α-epithio-17α-methyl-5α-androstan-17β-ol) is a designer steroid that is an ingredient in several "dietary" and "nutritional" supplements available online. As an unusual chemical modification to the steroid A-ring could allow this compound to pass through antidoping screens undetected, the metabolism of
Anna Panek et al.
Molecules (Basel, Switzerland), 25(18) (2020-09-19)
The biotransformation of steroid compounds is a promising, environmentally friendly route to new pharmaceuticals and hormones. One of the reaction types common in the metabolic fate of steroids is Baeyer-Villiger oxidation, which in the case of cyclic ketones, such as
Christopher D Chouinard et al.
Journal of the American Society for Mass Spectrometry, 28(2), 323-331 (2016-12-04)
Ion mobility-mass spectrometry (IM-MS) has recently seen increased use in the analysis of small molecules, especially in the field of metabolomics, for increased breadth of information and improved separation of isomers. In this study, steroid epimers androsterone and trans-androsterone were
Argitxu Esquivel et al.
Drug testing and analysis, 11(3), 392-402 (2018-10-27)
The detection of testosterone (T) misuse is performed using the steroid profile that includes concentrations of T and related metabolites excreted free and glucuronoconjugated, and the ratios between them. In this work, the usefulness of 14 endogenous steroid sulfates to
Seun F Akomolafe et al.
The Journal of pharmacy and pharmacology, 71(10), 1565-1575 (2019-08-07)
This study was designed to assess the role of caffeine on fertility parameters in testicular and epididymal tissues of scopolamine-induced model of amnesia in rats. Adult male rats were treated with scopolamine with or without caffeine. The modulatory effects of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service