Skip to Content
Merck
All Photos(1)

Key Documents

E1142

Sigma-Aldrich

β-Endorphin rat

≥97% (HPLC)

Synonym(s):

β-Lipotropin 61-91

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C157H254N42O44S
CAS Number:
Molecular Weight:
3466.02
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥97% (HPLC)

UniProt accession no.

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](NC(=O)[C@H](C)NC(=O)[C@H](CC(N)=O)NC(=O)[C@H](CCCCN)NC(=O)[C@H](Cc1ccccc1)NC(=O)[C@H](CC(C)C)NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@H](CC(C)C)NC(=O)[C@@H]2CCCN2C(=O)[C@@H](NC(=O)[C@H](CCC(N)=O)NC(=O)[C@H](CO)NC(=O)[C@H](CCCCN)NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](CO)NC(=O)[C@@H](NC(=O)[C@H](CCSC)NC(=O)[C@H](Cc3ccccc3)NC(=O)CNC(=O)CNC(=O)[C@@H](N)Cc4ccc(O)cc4)[C@@H](C)O)[C@@H](C)O)C(C)C)[C@@H](C)O)C(=O)N[C@@H]([C@@H](C)CC)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CC(N)=O)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](Cc5c[nH]cn5)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CCCCN)C(=O)NCC(=O)N[C@@H](CCC(N)=O)C(O)=O

InChI

1S/C157H254N42O44S/c1-18-84(11)125(152(237)183-100(45-30-35-62-162)135(220)186-111(72-118(167)209)146(231)192-123(82(7)8)150(235)189-109(70-93-73-168-79-172-93)144(229)179-97(42-27-32-59-159)133(218)176-96(41-26-31-58-158)132(217)171-76-121(212)174-104(157(242)243)52-55-116(165)207)195-153(238)126(85(12)19-2)194-130(215)86(13)173-140(225)110(71-117(166)208)185-134(219)98(43-28-33-60-160)178-143(228)108(69-91-39-24-21-25-40-91)184-141(226)105(65-80(3)4)188-154(239)128(88(15)203)197-151(236)124(83(9)10)193-145(230)106(66-81(5)6)187-149(234)114-46-36-63-199(114)156(241)129(89(16)204)198-138(223)101(51-54-115(164)206)180-147(232)112(77-200)190-136(221)99(44-29-34-61-161)177-137(222)102(53-56-122(213)214)181-148(233)113(78-201)191-155(240)127(87(14)202)196-139(224)103(57-64-244-17)182-142(227)107(68-90-37-22-20-23-38-90)175-120(211)75-169-119(210)74-170-131(216)95(163)67-92-47-49-94(205)50-48-92/h20-25,37-40,47-50,73,79-89,95-114,123-129,200-205H,18-19,26-36,41-46,51-72,74-78,158-163H2,1-17H3,(H2,164,206)(H2,165,207)(H2,166,208)(H2,167,209)(H,168,172)(H,169,210)(H,170,216)(H,171,217)(H,173,225)(H,174,212)(H,175,211)(H,176,218)(H,177,222)(H,178,228)(H,179,229)(H,180,232)(H,181,233)(H,182,227)(H,183,237)(H,184,226)(H,185,219)(H,186,220)(H,187,234)(H,188,239)(H,189,235)(H,190,221)(H,191,240)(H,192,231)(H,193,230)(H,194,215)(H,195,238)(H,196,224)(H,197,236)(H,198,223)(H,213,214)(H,242,243)/t84-,85-,86-,87+,88+,89+,95-,96-,97-,98-,99-,100-,101-,102-,103-,104-,105-,106-,107-,108-,109-,110-,111-,112-,113-,114-,123-,124-,125-,126-,127-,128-,129-/m0/s1

InChI key

KSZMZNRVPCEDEO-KFHPFRHLSA-N

Gene Information

rat ... Pomc(24664)

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Tyr-Gly-Gly-Phe-Met-Thr-Ser-Glu-Lys-Ser-Gln-Thr-Pro-Leu-Val-Thr-Leu-Phe-Lys-Asn-Ala-Ile-Ile-Lys-Asn-Val-His-Lys-Lys-Gly-Gln

Biochem/physiol Actions

Potent endogenous opioid protein that is derived from propiomelanocortin, a protein found in the brain, anterior pituitary, skin, immune system, and other peripheral sites. It is released in response to painful stimuli and has potent antinociceptive activity that is mediated through its action on μ receptors in brain and by μ and κ receptors in the spinal cord.

Other Notes

Lyophilized from 0.1% TFA in H2O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

N Wang et al.
Journal of thrombosis and haemostasis : JTH, 2(5), 785-796 (2004-04-22)
Plasminogen-deficient mice exhibit behavioral differences in response to stress, including a markedly reduced acoustic startle reflex response compared with wild-type (WT) littermates. The acoustic startle reflex activates the hypothalamic-pituitary axis and is modulated by these hormones. The purpose of this
Gil Myoung Kang et al.
Cell metabolism, 33(2), 334-349 (2021-02-04)
Low-grade mitochondrial stress can promote health and longevity, a phenomenon termed mitohormesis. Here, we demonstrate the opposing metabolic effects of low-level and high-level mitochondrial ribosomal (mitoribosomal) stress in hypothalamic proopiomelanocortin (POMC) neurons. POMC neuron-specific severe mitoribosomal stress due to Crif1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service