Skip to Content
Merck
All Photos(2)

Documents

D6920

Sigma-Aldrich

2′-Deoxyadenosine 5′-triphosphate sodium salt solution

10 mM

Synonym(s):

dATP

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41106305
NACRES:
NA.52

Assay

≥99%

Quality Level

form

liquid

concentration

10 mM

color

colorless

foreign activity

DNase, RNase, NICKase, none detected

shipped in

dry ice

storage temp.

−20°C

SMILES string

[Na+].Nc1ncnc2n(cnc12)[C@H]3C[C@H](O)[C@@H](COP(O)(=O)OP(O)(=O)OP(O)([O-])=O)O3

InChI

1S/C10H16N5O12P3.Na/c11-9-8-10(13-3-12-9)15(4-14-8)7-1-5(16)6(25-7)2-24-29(20,21)27-30(22,23)26-28(17,18)19;/h3-7,16H,1-2H2,(H,20,21)(H,22,23)(H2,11,12,13)(H2,17,18,19);/q;+1/p-1/t5-,6+,7+;/m0./s1

InChI key

YJWCICGGRLOGEH-VWZUFWLJSA-M

Looking for similar products? Visit Product Comparison Guide

General description

2′-Deoxyadenosine 5′-triphosphate (dATP) is made up of a nucleobase attached to deoxyribose and a 5′-hydroxyl on the sugar bound to a chain of three phosphate residues. dATP is used by cells for synthesis of DNA by DNA polymerases.

Application

2′-Deoxyadenosine 5′-triphosphate sodium salt solution has been used as a component of the dNTP mix for labelling of ribosomal 18s rDNA probe and telomeric (TTAGG)n probe.
2′-Deoxyadenosine 5′-triphosphate sodium salt solution is used in DNA sysnthesis reactions such as PCR, DNA sequencing and molecular cloning techniques.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Spittlebugs (Hemiptera)
Kuznetsova V G, et al.
Protocols for Cytogenetic Mapping of Arthropod Genomes., 351-380 (2015)
Serdal Kirmizialtin et al.
Structure (London, England : 1993), 20(4), 618-627 (2012-04-10)
Nearly every enzyme undergoes a significant change in structure after binding it's substrate. Experimental and theoretical analyses of the role of changes in HIV reverse transcriptase structure in selecting a correct substrate are presented. Atomically detailed simulations using the Milestoning
Kousuke Sato et al.
Chembiochem : a European journal of chemical biology, 12(15), 2341-2346 (2011-09-03)
We herein describe the synthesis of fluorescent 5-(5,6-dimethoxybenzothiazol-2-yl)-2'-deoxyuridine 5'-triphosphate (d(bt)UTP) and primer extension reactions using d(bt)UTP. We also carried out primer extension reactions using the (bt)U template. B family DNA polymerases, such as KOD, Deep Vent (exo-), and 9°N(m) DNA
U C Halder et al.
Cell death & disease, 2, e197-e197 (2011-09-02)
During early infection, viruses activate cellular stress-response proteins such as heat-shock proteins (Hsps) to counteract apoptosis, but later on, they modulate these proteins to stimulate apoptosis for efficient viral dissemination. Hsp70 has been attributed to modulate viral entry, transcription, nuclear
Christina M Zimanyi et al.
Structure (London, England : 1993), 20(8), 1374-1383 (2012-06-26)
Ribonucleotide reductases (RNRs) provide the precursors for DNA biosynthesis and repair and are successful targets for anticancer drugs such as clofarabine and gemcitabine. Recently, we reported that dATP inhibits E. coli class Ia RNR by driving formation of RNR subunits

Protocols

Protocol using antibody mediated hot start polymerase with a red dye for easy gel loading. Method has short activation period (<1min), and results in higher yields and more specificity over standard PCR methods.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service