Skip to Content
Merck
All Photos(4)

Key Documents

D5412

Sigma-Aldrich

2′-Deoxyuridine

≥98.5%

Synonym(s):

1-(2-Deoxy-β-D-ribofuranosyl)uracil, Uracil deoxyriboside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H12N2O5
CAS Number:
Molecular Weight:
228.20
Beilstein:
24433
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

biological source

synthetic (organic)

Assay

≥98.5%

form

powder

impurities

Thymidine, free

solubility

water: 50 mg/mL, clear to slightly hazy, colorless to faintly yellow

SMILES string

OC[C@H]1O[C@H](C[C@@H]1O)N2C=CC(=O)NC2=O

InChI

1S/C9H12N2O5/c12-4-6-5(13)3-8(16-6)11-2-1-7(14)10-9(11)15/h1-2,5-6,8,12-13H,3-4H2,(H,10,14,15)/t5-,6+,8+/m0/s1

InChI key

MXHRCPNRJAMMIM-SHYZEUOFSA-N

Gene Information

mouse ... Slc29a1(63959)

Looking for similar products? Visit Product Comparison Guide

Application

2′-Deoxyuridine has been used as a:
  • precursor of [3H]thymidine triphosphate (TTP) in place of thymidine to avoid a potential thymidine block in relative proliferation assays
  • nucleoside supplement for cell cycle synchronization and DNA replication inhibition
  • mitotic inhibitor in culture media to minimize the proliferation of glial cells

Biochem/physiol Actions

2′-Deoxyuridine (dU) is frequently halogenated to create thymidine analogs useful for studies of DNA synthesis and degradation mechanisms. Derivatized 2′-deoxyuridines used as labeling substrates include chloro-2′-deoxyuridine (CldU), bromodeoxyuridine (BrdU) and/or iododeoxyuridine (IdU). Other useful analogs of 2′-deoxyuridine include 5-ethynyl-2′-deoxyuridine (DdU) and 5-hydroxymethyl-2′-deoxyuridine (HmdU).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yang Yang et al.
Cancer letters, 453, 170-183 (2019-04-08)
Overexpression of NQO1 is associated with poor prognosis in human cancers including lung, stomach, colon, cervical, and pancreatic cancers. However, the molecular mechanisms underlying the protumorigenic capacities of NQO1 have not been fully elucidated. Here, we investigated this question and
Quan-Liang Jian et al.
PeerJ, 6, e5739-e5739 (2018-10-09)
Both age and intensive exercise are generally considered critical risk factors for osteoarthritis. In this work, we intend to establish zebrafish models to assess the role of these two factors on cartilage homeostasis. We designed a swimming device for zebrafish
Kaori Nomura-Komoike et al.
Scientific reports, 10(1), 1488-1488 (2020-02-01)
Müller glia, the principal glial cell type in the retina, have the potential to reenter the cell cycle after retinal injury. In mammals, proliferation of Müller glia is followed by gliosis, but not regeneration of neurons. Retinal injury is also
Hiroyuki Yamakoshi et al.
Journal of the American Chemical Society, 134(51), 20681-20689 (2012-12-04)
Alkyne has a unique Raman band that does not overlap with Raman scattering from any endogenous molecule in live cells. Here, we show that alkyne-tag Raman imaging (ATRI) is a promising approach for visualizing nonimmobilized small molecules in live cells.
Hongxiu Ning et al.
Cytotherapy, 15(1), 57-63 (2012-12-25)
Thymidine analog 5-ethynyl-2-deoxyuridine (EdU) has recently been used for tracking mesenchymal stem cells (MSCs). In the present study, we tested whether EdU was cytotoxic and whether it interfered with differentiation, cytokine secretion and migration of MSCs. EdU labeling was performed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service