Skip to Content
Merck
All Photos(1)

Key Documents

D2779

Sigma-Aldrich

Dimethyldioctadecylammonium bromide

≥98% (TLC)

Synonym(s):

Distearyldimethylammonium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)17]2N(Br)(CH3)2
CAS Number:
Molecular Weight:
630.95
Beilstein:
3580323
EC Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.25

description

cationic

Assay

≥98% (TLC)

form

powder

mol wt

630.95 g/mol

mp

~160 °C

application(s)

cell analysis

SMILES string

CCCCCCCCCCCCCCCCCC[N+](CCCCCCCCCCCCCCCCCC)(C)C.[Br-]

InChI

1S/C38H80N.BrH/c1-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35-37-39(3,4)38-36-34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-2;/h5-38H2,1-4H3;1H/q+1;/p-1

InChI key

PSLWZOIUBRXAQW-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Dimethyldioctadecylammonium bromide is a cationic lipid that stimulates immune responses, activates macrophages, and binds to antigens.

Application

Dimethyldioctadecylammonium bromide has been used in a study to assess the interaction of its monolayers with DNA using specular neutron reflection, surface pressure-area isotherms, and Brewster angle microscopy. It has also been used in a study to investigate new adjuvants for use in kill vaccines for bovine viral diarrhea virus (BVDV).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julia F Ridpath et al.
Veterinary research communications, 34(8), 691-702 (2010-09-28)
Bovine viral diarrhea virus (BVDV) infections cause respiratory, reproductive, and enteric disease in cattle. Vaccination raises herd resistance and limits the spread of BVDV among cattle. Both killed and modified live vaccines against BVDV are available. While modified live vaccines
F S Baechtel et al.
Cancer research, 42(12), 4959-4963 (1982-12-01)
Dimethyldioctadecylammonium bromide (DDA) stimulates immune responses, primes (or activates) macrophages, and binds to antigens. Because relatively little is known about the binding of adjuvants to antigens, the nature of the interaction of DDA with soluble protein and cellular antigens was
Eloi Feitosa et al.
PloS one, 7(9), e44702-e44702 (2012-09-13)
Dioctadecyldimethylammonium bromide (DODAB) is a double chain cationic lipid, which assembles as bilayer structures in aqueous solution. The precise structures formed depend on, e.g., lipid concentration and temperature. We here combine differential scanning calorimetry (DSC) and X-ray scattering (SAXS and
Agnes Michanek et al.
Langmuir : the ACS journal of surfaces and colloids, 28(25), 9621-9633 (2012-05-26)
The objective of this work is to establish under which conditions short RNA molecules (similar to miRNA) associate with zwitterionic phospholipids and how this differs from the association with cationic surfactants. We study how the base pairing (i.e., single stranded
Paolo Sabatino et al.
The journal of physical chemistry. B, 116(30), 9269-9276 (2012-06-27)
The interaction between phenol molecules, both in their undissociated and dissociated states, and cationic dioctadecyl dimethylammonium chloride (DODAC) vesicles were thoroughly investigated using NMR techniques. In particular, diffusion and relaxation measurements, combined with the two sites Kärger model, were used

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service