Skip to Content
Merck
All Photos(4)

Documents

D1692

Sigma-Aldrich

GW4869

≥90% (NMR), powder, N-SMase inhibitor

Synonym(s):

N,N′-Bis[4-(4,5-dihydro-1H-imidazol-2-yl)phenyl]-3,3′-p-phenylene-bis-acrylamide dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H28N6O2 · 2HCl
CAS Number:
Molecular Weight:
577.50
UNSPSC Code:
41121800
PubChem Substance ID:
NACRES:
NA.77

product name

GW4869, ≥90% (NMR)

Quality Level

Assay

≥90% (NMR)

form

powder

storage condition

desiccated
protect from light

color

light yellow to yellow

mp

>300 °C

solubility

DMSO: 0.2 mg/mL

originator

GlaxoSmithKline

storage temp.

2-8°C

SMILES string

Cl.Cl.O=C(Nc1ccc(cc1)C2=NCCN2)\C=C/c3ccc(\C=C/C(=O)Nc4ccc(cc4)C5=NCCN5)cc3

InChI

1S/C30H28N6O2.2ClH/c37-27(35-25-11-7-23(8-12-25)29-31-17-18-32-29)15-5-21-1-2-22(4-3-21)6-16-28(38)36-26-13-9-24(10-14-26)30-33-19-20-34-30;;/h1-16H,17-20H2,(H,31,32)(H,33,34)(H,35,37)(H,36,38);2*1H/b15-5-,16-6-;;

InChI key

NSFKAZDTKIKLKT-LOLTXFFGSA-N

General description

GW4869 is a commonly used pharmacological agent, which inhibits exosome generation. It blocks ceramide-mediated inward budding of multivesicular bodies (MVBs) and the release of mature exosomes from MVBs. GW4869 exhibits cytotoxicity to phosphatidylserine-expressing myeloma cells. It inhibits the secretion of IFN (interferon)-α by plasmacytoid dendritic cells (pDCs).

Application

GW4869 has been used:
  • as an inhibitor of neutral sphingomyelinase and exosome biogenesis
  • to analyse the effects of arsenic trioxide (ATO) treatment for hepatoma carcinoma HCCLM3 cells on ceramide production
  • to determine the contributions of p75 neurotrophin receptor (p75NTR) and tropomyosin receptor kinase A (TrkA)- coupled pathways to nerve growth factor (NGF)-induced thermal hypersensitivity in rats

Biochem/physiol Actions

A cell-permeable, potent, specific, non-competitive inhibitor of N-SMase (neutral sphingomyelinase)

Features and Benefits

This compound was developed by GlaxoSmithKline. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The cationic small molecule GW 4869 is cytotoxic to high phosphatidylserine-expressing myeloma cells
Vuckovic S, et al.
British Journal of Haematology, 177(3), 423-440 (2017)
Elevated Wall Tension Leads to Reduced miR-133a in the Thoracic Aorta by Exosome Release
Akerman AW, et al.
Journal of the American Heart Association, 8(1), e010332-e010332 (2019)
The TrkA receptor mediates experimental thermal hyperalgesia produced by nerve growth factor: modulation by the p75 neurotrophin receptor
Khodorova A, et al.
Neuroscience, 340, 384-397 (2017)
Phillip B Munson et al.
Scientific reports, 9(1), 11688-11688 (2019-08-14)
Malignant mesothelioma (MM) is an asbestos-induced cancer arising on the mesothelial surface of organ cavities. MM is essentially incurable without a means of early diagnosis and no successful standard of care. These facts indicate a deep chasm of knowledge that
Yunlong Huang et al.
Cell discovery, 4, 19-19 (2018-05-01)
Zika virus (ZIKV) is a neurotrophic flavivirus that is capable of infecting humans, leading to brain abnormalities during fetal development. The ZIKV infectivity in neural target cells remains poorly understood. Here, we found that ZIKV specifically infected glial fibrillary acidic

Articles

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service