Skip to Content
Merck
All Photos(1)

Key Documents

D1641

Sigma-Aldrich

Dihydrocytochalasin B

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H39NO5
CAS Number:
Molecular Weight:
481.62
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.32

form

powder

Quality Level

storage temp.

−20°C

SMILES string

C[C@@H]1CCC[C@@H](O)CCC(=O)O[C@]23[C@@H](\C=C\C1)[C@H](O)C(=C)[C@@H](C)[C@H]2[C@H](Cc4ccccc4)NC3=O

InChI

1S/C29H39NO5/c1-18-9-7-13-22(31)15-16-25(32)35-29-23(14-8-10-18)27(33)20(3)19(2)26(29)24(30-28(29)34)17-21-11-5-4-6-12-21/h4-6,8,11-12,14,18-19,22-24,26-27,31,33H,3,7,9-10,13,15-17H2,1-2H3,(H,30,34)/b14-8+/t18-,19-,22-,23+,24+,26+,27-,29-/m1/s1

InChI key

WIULKAASLBZREV-RXPQEOCGSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Marker for high affinity cytochalasin B binding sites.

Biochem/physiol Actions

Effect on cell motility and morphology similar to that of cytochalasin B; does not inhibit glucose transport.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Carc. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P J Rijken et al.
Experimental cell research, 243(2), 254-262 (1998-09-23)
Epidermal growth factor (EGF) induces rapid actin filament assembly in the membrane skeleton of a variety of cells. To investigate the significance of this process for signal transduction, actin polymerization is inhibited by dihydrocytochalasin B (CB). CB almost completely abolishes
Lucia Formigli et al.
Journal of cellular physiology, 211(2), 296-306 (2007-02-14)
In the present study, we investigated the functional interaction between stress fibers (SFs) and stretch-activated channels (SACs) and its possible role in the regulation of myoblast differentiation induced by switch to differentiation culture in the presence or absence of sphingosine
A Scherberich et al.
Journal of cell science, 113 ( Pt 4), 653-662 (2000-02-01)
Migration of vascular smooth muscle cells (VSMC) is a key event in the formation of neointima during atherosclerosis. Fura-2 loaded VSMCs were used to investigate calcium homeostasis during cell migration. Multiple spontaneous transient increases in cytosolic free calcium [Ca(2+)](i)were observed
Olivier D Lohez et al.
The Journal of cell biology, 161(1), 67-77 (2003-04-16)
p53 and the retinoblastoma (RB) pocket proteins are central to the control of progression through the G1 phase of the cell cycle. The RB pocket protein family is downstream of p53 and controls S-phase entry. Disruption of actin assembly arrests
P R Andreassen et al.
Mutation research, 372(2), 181-194 (1996-12-01)
Populations of tetraploid cells are found in a variety of human tumours where they may act as precursors of aneuploidy and tumorigenesis. Here we demonstrate the drug induction of tetraploid cells at mitosis by interference with cell cycle checkpoints and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service