Skip to Content
Merck
All Photos(1)

Key Documents

D1262

Sigma-Aldrich

P1,P4-Di(adenosine-5′) tetraphosphate ammonium salt

≥95% (HPLC), powder

Synonym(s):

A(5′)p4(5′)A, Diadenosine tetraphosphate ammonium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H28N10O19P4 · xNH3
CAS Number:
Molecular Weight:
836.39 (free acid basis)
MDL number:
UNSPSC Code:
41106305
eCl@ss:
32160414
PubChem Substance ID:
NACRES:
NA.77

Assay

≥95% (HPLC)

form

powder

color

white to off-white

solubility

H2O: 50 mg/mL

storage temp.

−20°C

SMILES string

N.Nc1ncnc2n(cnc12)[C@@H]3O[C@H](COP(O)(=O)OP(O)(=O)OP(O)(=O)OP(O)(=O)OC[C@H]4O[C@H]([C@H](O)[C@@H]4O)n5cnc6c(N)ncnc56)[C@@H](O)[C@H]3O

InChI

1S/C20H28N10O19P4.H3N/c21-15-9-17(25-3-23-15)29(5-27-9)19-13(33)11(31)7(45-19)1-43-50(35,36)47-52(39,40)49-53(41,42)48-51(37,38)44-2-8-12(32)14(34)20(46-8)30-6-28-10-16(22)24-4-26-18(10)30;/h3-8,11-14,19-20,31-34H,1-2H2,(H,35,36)(H,37,38)(H,39,40)(H,41,42)(H2,21,23,25)(H2,22,24,26);1H3/t7-,8-,11-,12-,13-,14-,19-,20-;/m1./s1

InChI key

JTLRVYSRZSPJKJ-AHKIGRPSSA-N

Application

P1,P4-Di(adenosine-5′) tetraphosphate ammonium salt has been used as a substrate for nudix hydrolyse. It has also been used as a ATP analog and nudix substrate for cleavage factor II Im25 subunit.

Biochem/physiol Actions

Diadenosine polyphosphate (Ap4A) functions as a second messenger in pancreatic cells. Diadenosine polyphosphate is majorly stored in neuronal cells, thrombocytes and chromaffin. It stimulates phospholipase D, fluctuates intracellular calcium levels, induces nitric oxide release, activates 5′-nucleotidase, and inhibits adenosine kinase activity in vitro. The membrane-bound ectoenzymes in endothelial cells in blood plasma and smooth muscle cell metabolize Ap4A.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adenine dinucleotides: a novel class of signalling molecules.
Ogilvie A, et al.
Journal of Autonomic Pharmacology, 16(6), 325-328 (1996)
Diadenosine polyphosphates in insulin-secreting cells: interaction with specific receptors and degradation.
Verspohl EJ and Johannwille B
Diabetes, 47(11), 1727-1734 (1998)
S S Wildman et al.
European journal of pharmacology, 367(1), 119-123 (1999-03-19)
The pharmacological activity of diadenosine polyphosphates was investigated at three recombinant P2X receptors (rat P2X1, rat P2X3, rat P2X4) expressed in Xenopus oocytes and studied under voltage-clamp conditions. For the rat P2X1 receptor, only P1,P6-diadenosine hexaphosphate (Ap6A) was a full
The Ralstonia solanacearum effector RipN suppresses plant PAMP-triggered immunity, localizes to the endoplasmic reticulum and nucleus, and alters the NADH/NAD+ ratio in Arabidopsis
Sun Y, et al.
Molecular Plant Pathology, 20(4), 533-546 (2019)
R H Hilderman et al.
FEBS letters, 427(3), 320-324 (1998-06-24)
Since the infusion of P1,P4-diadenosine 5' tetraphosphate (Ap4A) into animal models induces vasodilation [1,2], the present study was performed to determine whether Ap4A induces the release of nitric oxide (NO) from endothelial cells. Ap4A induced NO release was 4.2-fold greater

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service