Skip to Content
Merck

Skip To

D101

(±)-DOI hydrochloride

≥98% (HPLC), solid,  5-HT2 serotonin receptor agonist

Synonym(s):

(±)-1-(2,5-Dimethoxy-4-iodophenyl)-2-aminopropane hydrochloride, (±)-2,5-Dimethoxy-4-iodoamphetamine hydrochloride, (±)-DOI

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C11H16INO2 · HCl
CAS Number:
Molecular Weight:
357.62
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
solid
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

(±)-DOI hydrochloride, ≥98% (HPLC), solid

Quality Segment

assay

≥98% (HPLC)

form

solid

drug control

Home Office Schedule 1; regulated under CDSA - not available from Sigma-Aldrich Canada, kontrollierte Droge in Deutschland

color

white to off-white

solubility

H2O: 10 mg/mL, ethanol: 2 mg/mL

SMILES string

NC(C)CC1=CC(OC)=C(I)C=C1OC.Cl

InChI

1S/C11H16INO2.ClH/c1-7(13)4-8-5-11(15-3)9(12)6-10(8)14-2;/h5-7H,4,13H2,1-3H3;1H

InChI key

QVFDMWGKHUFODK-UHFFFAOYSA-N

Gene Information

Biochem/physiol Actions

(±)-DOI hydrochloride is a potent and selective 5-HT2 serotonin receptor agonist that crosses the blood-brain barrier, increases dopamine turnover and the treatment with DOI counteracted the expression of the nicotine-induced locomotor and neurochemical sensitization, but had no effect on nicotine-induced behavioral disinhibition. The responses to DOI provide evidence that of these receptors are abundant in the cortex, and show relatively lower levels in hippocampus and cerebellum.

Legal Information

German
Dieses Produkt fällt unter das Betäubungsmittelgesetz (BtMG). Für eine Bestellung dieses Produktes ist eine Erlaubnis nach § 3 BtMG zwingend erforderlich, es sei denn, es greift eine Ausnahme von der Erlaubnispflicht nach § 4 oder § 26 BtMG.

English
This product is subject to the German Narcotics Act. A permit under Section 3 of the German Narcotics Act is mandatory for ordering this product unless an exemption from the permit requirement under Section 4 or Section 26 of the German Narcotics Act applies.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
D153G1421D6821
form

solid

form

solid

form

solid

form

powder

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

drug control

Home Office Schedule 1; regulated under CDSA - not available from Sigma-Aldrich Canada, kontrollierte Droge in Deutschland

drug control

Home Office Schedule 1; regulated under CDSA - not available from Sigma-Aldrich Canada, kontrollierte Droge in Deutschland

drug control

-

drug control

-

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

solubility

H2O: 10 mg/mL, ethanol: 2 mg/mL

solubility

H2O: ≥20 mg/mL, ethanol: soluble

solubility

DMSO: soluble >10 mg/mL, H2O: insoluble <2 mg/mL

solubility

DMSO: ≥20 mg/mL, H2O: ≥20 mg/mL

color

white to off-white

color

white

color

white to pink

color

white to off-white


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters