All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C122H223N53O23 · xC2HF3O2
Molecular Weight:
2800.42 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77
Recommended Products
Assay
≥95% (HPLC)
form
film
composition
Peptide content, ≥60%
color
colorless to white
solubility
H2O: 0.5 mg/mL, clear
shipped in
wet ice
storage temp.
−20°C
Amino Acid Sequence
Tyr-Gly-Arg-Lys-Lys-Arg-Arg-Gln-Arg-Arg-Arg-Pro-Pro-Leu-Arg-Lys-Lys-Lys-Lys-Lys-His-NH2
Biochem/physiol Actions
Cell permeable cGMP protein kinase (PKG) inhibitor
Features and Benefits
This compound is featured on the PKA & PKG page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Mark S Taylor et al.
Molecular pharmacology, 65(5), 1111-1119 (2004-04-23)
Cyclic GMP-dependent protein kinase (PKG) serves as an important physiological regulator of vascular reactivity and tone. However, available inhibitors of PKG have exhibited variable effects in intact tissue, hindering the elucidation of the functional role of PKG in blood vessels.
W R Dostmann et al.
Proceedings of the National Academy of Sciences of the United States of America, 97(26), 14772-14777 (2000-12-20)
Arrays of octameric peptide libraries on cellulose paper were screened by using (32)P-autophosphorylated cGMP-dependent protein kinase Ialpha (cGPK) to identify peptide sequences with high binding affinity for cGPK. Iterative deconvolution of every amino acid position in the peptides identified the
Christin Christensen et al.
Stem cells and development, 15(5), 647-654 (2006-11-16)
Previously, we have shown that increasing the intracellular cGMP concentration enhances the sonic hedgehog (Shh) response in neural plate cells. The use of two mouse embryonic stem (ES) cell lines allowed a highly sensitive and reproducible quantification of the Shh
Articles
Our offers many products related to PKA & PKG for your research needs.
Our offers many products related to PKA & PKG for your research needs.
Our offers many products related to PKA & PKG for your research needs.
Our offers many products related to PKA & PKG for your research needs.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service