Skip to Content
Merck
All Photos(2)

Documents

D0138

Sigma-Aldrich

1,2-Dioleoyl-sn-glycerol

≥97%

Synonym(s):

(9Z)-9-Octadecenoic acid 1,1′-[(1S)-1-(hydroxymethyl)-1,2-ethanediyl] ester, (S)-1,2-Diolein, (S)-Glycerol 1,2-dioleate, 1,2-Di(cis-9-octadecenoyl)-sn-glycerol, DG(18:1(9Z)/18:1(9Z)/0:0), DG(18:1/18:1/0:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H72O5
CAS Number:
Molecular Weight:
620.99
Beilstein:
1730457
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

biological source

semisynthetic

Quality Level

Assay

≥97%

form

liquid

impurities

1,3-isomer, trace

functional group

ether

lipid type

neutral glycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC[C@H](OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O

InChI

1S/C39H72O5/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-38(41)43-36-37(35-40)44-39(42)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h17-20,37,40H,3-16,21-36H2,1-2H3/b19-17-,20-18-/t37-/m0/s1

InChI key

AFSHUZFNMVJNKX-LLWMBOQKSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Direct and simultaneous analysis of lipase-catalyzed hydrolysis of high-oleic oil model by chiral stationary phase HPLC-ELSD.: This study utilizes 1,2-dioleoyl-sn-glycerol to investigate lipase-catalyzed hydrolysis, providing insights into enzymatic processes and chiral analysis which are critical in food chemistry and biochemistry (Choi et al., 2022).

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hanako Nishikawa et al.
Bio-protocol, 10(10), e3626-e3626 (2021-03-05)
It is important to experimentally determine how membrane proteins are integrated into biomembranes to unveil the roles of the integration factors, and to understand the functions and structures of membrane proteins. We have developed a reconstitution system for membrane protein
Peter Reinink et al.
European journal of immunology, 49(5), 737-746 (2019-03-12)
Lyme disease is a common multisystem disease caused by infection with a tick-transmitted spirochete, Borrelia burgdorferi and related Borrelia species. The monoglycosylated diacylglycerol known as B. burgdorferi glycolipid II (BbGL-II) is a major target of antibodies in sera from infected
Rachel N Cotton et al.
The Journal of clinical investigation, 131(1) (2021-01-05)
CD1a-autoreactive T cells contribute to skin disease, but the identity of immunodominant self-lipid antigens and their mode of recognition are not yet solved. In most models, MHC and CD1 proteins serve as display platforms for smaller antigens. Here, we showed
Ferdinand Ngale Njume et al.
PLoS neglected tropical diseases, 13(7), e0007591-e0007591 (2019-07-23)
Onchocerca volvulus is the nematode pathogen responsible for human onchocerciasis also known as "River blindness", a neglected tropical disease that affects up to 18 million people worldwide. Helminths Excretory Secretory Products (ESPs) constitute a rich repertoire of molecules that can
Keisuke Kobayashi et al.
Scientific reports, 8(1), 12099-12099 (2018-08-16)
Atropisomers with a biaryl dihydronaphthopyranone structure, dinapinones A1 (DPA1) (M position) and A2 (DPA2) (P position), were isolated from the fungus culture broth of Talaromyces pinophilus FKI-3864 as inhibitors of [14C]neutral lipid ([14C]triacylglycerol (TG) and [14C]cholesteryl ester (CE)) synthesis from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service