Skip to Content
Merck
All Photos(1)

Key Documents

CYTOCOX1

Sigma-Aldrich

Cytochrome c Oxidase Assay Kit

sufficient for 100 tests, soluble and membrane bound mitochondria

Synonym(s):

Cytochrome c Oxidase Activity Assay Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

Quality Level

usage

sufficient for 100 tests

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... COX1(4512)

Application

The Cytochrome c Oxidase Assay Kit uses an optimized colorimetric assay based on observation of the decrease in absorbance of ferrocytochrome c measured at 550 nm, which is caused by its oxidation to ferricytochrome c by cytochrome c oxidase. This kit is suitable for the detection of mitochondrial outer membrane integrity/mitochondrial stress and for the detection of mitochondria in subcellular fractions.

Biochem/physiol Actions

Cytochrome c oxidase [EC 1.9.3.1] is located on the inner mitochondrial membrane dividing the mitochondrial matrix from the intermembrane space, and has traditionally been used as a marker for this membrane. It is also located in the cytoplasmic membrane of bacteria. Cytochrome c oxidase provides energy for the cell by coupling electron transport through the cytochrome chain with the process of oxidative phosphorylation.

Features and Benefits

  • Simple, optimized protocol - Obtain reproducible results without special training needs
  • Useful for determining cytochrome c activity from any mitchondrial source - The enzyme is present in all mitochondria regardless of species
  • Useful for detecting the presence of mitochondria in subcellular fraction - Save time and increase confidence in the quality of organelle preparations
  • May be used in conjunction with the MITOISO1 Mitochondrial Isolation Kit - Standardized mitochondrial preparation and analysis ensures reproducibility
  • Use to analyze the intactness of mitochondrial membranes
  • Simple colorimetric measurement of solution

Kit Components Also Available Separately

Product No.
Description
SDS

  • C2506Cytochrome c from equine heart, ≥95% (SDS-PAGE) 50 mgSDS

  • D4641n-Dodecyl β-D-maltoside, ≥98% (GC) 10 mgSDS

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chao-Chin Liu et al.
Frontiers in microbiology, 11, 170-170 (2020-03-03)
Staphylococcus aureus spreads rapidly on the surface of soft agar medium. The spreading depends on the synthesis of biosurfactants, i.e., phenol soluble modulins (PSMs), which facilitate colony spreading of S. aureus. Our earlier study demonstrated that water accumulates in a
Céline Aguer et al.
PloS one, 6(12), e28536-e28536 (2011-12-24)
Human primary myotubes are highly glycolytic when cultured in high glucose medium rendering it difficult to study mitochondrial dysfunction. Galactose is known to enhance mitochondrial metabolism and could be an excellent model to study mitochondrial dysfunction in human primary myotubes.
Filip J Larsen et al.
Cell metabolism, 13(2), 149-159 (2011-02-03)
Nitrate, an inorganic anion abundant in vegetables, is converted in vivo to bioactive nitrogen oxides including NO. We recently demonstrated that dietary nitrate reduces oxygen cost during physical exercise, but the mechanism remains unknown. In a double-blind crossover trial we
Filip J Larsen et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 25(8), 2843-2852 (2011-05-18)
The basal metabolic rate (BMR) is referred to as the minimal rate of metabolism required to support basic body functions. It is well known that individual BMR varies greatly, even when correcting for body weight, fat content, and thyroid hormone
Uta Keil et al.
The Journal of biological chemistry, 279(48), 50310-50320 (2004-09-17)
Increasing evidence suggests an important role of mitochondrial dysfunction in the pathogenesis of Alzheimer's disease. Thus, we investigated the effects of acute and chronic exposure to increasing concentrations of amyloid beta (Abeta) on mitochondrial function and nitric oxide (NO) production

Articles

Centrifugation separates organelles based on size, shape, and density, facilitating subcellular fractionation across various samples.

Centrifugation separates organelles based on size, shape, and density, facilitating subcellular fractionation across various samples.

Centrifugation separates organelles based on size, shape, and density, facilitating subcellular fractionation across various samples.

Centrifugation separates organelles based on size, shape, and density, facilitating subcellular fractionation across various samples.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service