CLS430167
Corning® tissue-culture treated culture dishes
D × H 100 mm × 20 mm
Synonym(s):
petri dish
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121812
NACRES:
NA.75
Recommended Products
material
polystyrene
round
sterility
sterile
usage
11.0-16.5 mL (Optimal culture medium volume)
packaging
sleeve of 20
manufacturer/tradename
Corning 430167
D × H
100 mm × 20 mm
surface area
55 cm2
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Corning® Tissue Culture (TC)-treated items undergo surface modification through processes like corona discharge or gas plasma treatment. This modification leaves the surface hydrophilic and negatively charged with the addition of media.
Application
Corning®tissue-culture treated culture dishes are suitable for everyday attachment cell culture needs.
Features and Benefits
- Manufactured from optically clear virgin polystyrene
- Treated for optimal cell attachment
- Sterilized by gamma radiation and certified nonpyrogenic
- Have stacking beads to aid in handling
- Lid is supplied with vents to provide consistent gas exchange
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Sarika Saraswati et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 34(6), 7885-7904 (2020-04-17)
Nearly 6 million Americans suffer from heart failure. Increased fibrosis contributes to functional decline of the heart that leads to heart failure. Previously, we identified a mechanosensitive protein, small proline-rich repeat 3 (SPRR3), in vascular smooth muscle cells of atheromas.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service