CLS353181
Corning® Falcon® Permeable Support
for use with 12 Well Plate, with 3 µm Transparent PET Membrane, sterile
Synonym(s):
Permeable Support
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.13
Recommended Products
sterility
sterile
feature
with 3 µm Transparent PET Membrane
packaging
case of 48 ea (Individually wrapped)
manufacturer/tradename
Corning 353181
compatibility
for use with 12 Well Plate
Looking for similar products? Visit Product Comparison Guide
General description
Falcon® Permeable Support for 12 Well Plate with 3.0µm Transparent Polyester (PET) Membrane, Sterile, 1/Pack, 48/Case
Application
Cell Culture
Legal Information
Corning is a registered trademark of Corning, Inc.
Falcon is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service