Skip to Content
Merck
All Photos(1)

Key Documents

CLS3237

Corning® CellCube® Tubing Adaptor

3/8”ID Thermoelastic Tubing, Aseptic Connector to Female MPC, sterile

Synonym(s):

flask accessory

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122104
NACRES:
NB.13

material

polystyrene
tubing (3/8”ID Thermoelastic)

sterility

sterile

packaging

case of 4 ea

manufacturer/tradename

Corning 3237

Looking for similar products? Visit Product Comparison Guide

General description

  • CellCube Adaptor
  • Aseptic Connector to Female MPC

Application

Adherent Cell Scale Up

Legal Information

CELLCUBE is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service