Skip to Content
Merck
All Photos(2)

Key Documents

CLS20037

Corning® HYPERStack® 36-layer cell culture vessel

non-treated surface, sterile, bag of 1 ea, case of 2 ea

Synonym(s):

Corning container

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122104
NACRES:
NB.13

material

PTFE filters (membrane)
polycarbonate filling tube
polycarbonate plug (membrane)
polystyrene body (virgin)
polystyrene film
rectangular flask

sterility

sterile

packaging

bag of 1 ea
case of 2 ea

manufacturer/tradename

Corning 20037

technique(s)

cell culture | mammalian: suitable

surface area

18000 cm2 , cell growth area

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

The utilization of gas-permeable film technology allows Corning® High Yield PERformance (HYPER) cell culture vessels to provide more growth surface area versus traditional cell culture vessels of comparable footprint. With closed system accessories and multiple size offerings, Corning HYPERStack® cell culture vessels are an efficient, scalable cell culture vessel platform for adherent cell culture and can be used as seed train for adherent cell culture bioreactors.

Application

Adherent cell culture, cell expansion, stem cell expansion, cell banking, cell production, viral vector production, vaccine production

Features and Benefits

  • New updated design
  • Features 18,000 cm2 cell growth area
  • Corning High Yield PERformance (HYPER) gas-permeable film technology
  • Closed system approach with chemically resistant, heat sealable, flexible tubing and female MPC for liquid handling manipulations
  • Non-treated surface for ease of cell dissociation and harvest
  • Manfactured from USP Class VI virgin polystyrene which provides excellent optical clarity and mechanical strength
  • Sterilized to a sterility assurance level (SAL) 10-6
  • 100% leak tested throughout the manufacturing process

Legal Information

Corning is a registered trademark of Corning, Inc.
HYPERStack is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service