Recommended Products
Product Name
BE(2)-C, 95011817
biological source
human nerve
packaging
tube of 5 μg 95011817-DNA-5UG
pkg of vial of cells 95011817-1VL
growth mode
Adherent
karyotype
Modal no. 44
morphology
Neuroblast-like
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
metastasis
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Human Caucasian neuroblastoma
Cell Line Description
BE(2)-C is a clonal sub-line of SK-N-BE(2) (ECCAC catalogue no. 95011815) which was isolated from bone marrow of a 22-month-old male with disseminated neuroblastoma in 1972. They are reported to be multipotential with regard to neuronal enzyme expression and display a high capacity to convert tyrosine to dopamine. The cells show a small, refractile morphology with short, neurite-like cell processes and tend to grow in aggregates.The Y chromosome could not be detected in this cell line by short tandem repeat (STR)-PCR analysis when tested at ECACC. It is a known phenomenon that due to the increased genetic instability of cancer cell lines the Y chromosome can be rearranged or lost resulting in lack of detection. The cell line is identical to the source provided by the depositor based on the STR-PCR analysis.
Application
Chemotherapy development, chromosome studies, drug resistance, differentiation studies.
DNA Profile
STR-PCR Data: Amelogenin: X
CSF1PO: 10
D13S317: 11
D16S539: 9,11
D5S818: 12
D7S820: 9,10
THO1: 6
TPOX: 11
vWA: 18
CSF1PO: 10
D13S317: 11
D16S539: 9,11
D5S818: 12
D7S820: 9,10
THO1: 6
TPOX: 11
vWA: 18
Culture Medium
EMEM (EBSS) + 1% Non Essential Amino Acids (NEAA) : Ham′s F12 (1:1) + 2mM Glutamine + 15% Foetal Bovine Serum (FBS) (Heat Inactivated).
Subculture Routine
Split sub-confluent cultures (70-80%) 1:50 to 1:100 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service