IC-09
Reactivity: Anti-Oligodendrocytes and Schwann cells, bovine, 93062303
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
Mouse x Mouse Hybridoma
ImmunoDon
BALB/c mouse spleen
Myeloma
P3-NS1/1-Ag4.1
Reactivity
Anti-Oligodendrocytes and Schwann cells, bovine
technique(s)
cell culture | mammalian: suitable
isotype
IgG3
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Mouse P3-NS1/1-Ag4.1 x Mouse BALB/c spleen
Cell Line Description
The antibody is directed against heat-resistant and trypsin-resistant antigen present on bovine oligodendrocytes and Schwann cells and can be used in developmental studies of these cell types. All IC-09+ are also positive for IC-08 and galactocerebroside.
Immunogen
Particulate fraction from bovine corpus callosum
Application
Developmental studies and immunohistology
Culture Medium
DMEM + 2mM glutamine + 10% Foetal Bovine Serum (FBS)
Subculture Routine
Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service